Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546589_m1
          See other PPM1M GE Assays ›
          Gene Symbol:
          Ppm1m
          Gene Name:
          protein phosphatase 1M
          Gene Aliases:
          2810423O19Rik, AW610647, C77250, PP2Ceta
          Chromosome Location:
          Chr.9: 106194953 - 106199233 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546589_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppm1m
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026447.4 NP_080723.3 4 - 5 761 68
          NM_198931.3 NP_945149.2 4 - 5 765 68
          GenBank mRNA AB474373.1 - 4 - 5 800 68
          AK013149.1 - 4 - 5 694 68
          AK017245.1 - 1 - 2 98 68
          AK046387.1 - 660 68
          AK087999.1 - 4 - 5 799 68
          AY332616.1 - 4 - 5 768 68
          BC058248.1 - 4 - 5 687 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003003.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppm1m

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          catalytic activity
          phosphoprotein phosphatase activity
          CTD phosphatase activity
          hydrolase activity
          manganese ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801799 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801798 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801825 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801802 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801801 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801800 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033510 BC058248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033510 AK046387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033510 AK087999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033510 NM_198931.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033510 AB474373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138075 AK046387.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726201 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149264 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118099 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033510 BC058248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033510 AK046387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033510 AK087999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033510 NM_198931.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033510 AB474373.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P252545 BC058248.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P252545 AK046387.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P252545 NM_198931.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P252545 AK013149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P252545 AK017245.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26801818 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118099 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06914261 - -
          Whole Mouse Genome G4122A A_51_P252545 BC058248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P252545 AK046387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P252545 NM_198931.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P252545 AK013149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P252545 AK017245.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130499 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118099 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08098873 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08110684 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07672962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08110686 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07672963 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07672950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07672953 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07672948 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08098874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801802 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801799 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801798 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801801 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801825 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26801800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26801831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07672961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07672950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07524534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07524539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07995542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07524516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07524548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07672962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26801790 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10596451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596446 - -
          Affymetrix HT MG-430 PM Array Plate 1428351_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10596442 - -
          Mouse Genome 430 2.0 Array 1428351_at - -
          Mouse Genome 430 2.0 Array 1428351_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67905_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67905_at - -
          Affymetrix Mouse Clariom S Array PSR0900024923.mm.1 NM_026447.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024923.mm.1 NM_026447.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024923.mm.1 NM_198931.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024923.mm.1 NM_198931.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024923.mm.1 TC0900003003.mm.1 MTA TC
          Mouse Expression Set 430 1428351_PM_at - -
          Mouse Expression Set 430 1428351_at - -
          Murine Genome U74v2 Set 104525_at - -
          Murine Genome U74v2 Set 161992_PM_at - -
          Murine Genome U74v2 Set 161992_at - -
          Murine Genome U74v2 Set 104525_PM_at - -
          Mouse Genome 430A 2.0 Array 1428351_PM_at - -
          Mouse Genome 430A 2.0 Array 1428351_at - -
          Affymetrix HT Mouse Genome 430A Array 1428351_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428351_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 847357 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5270279 - -
          Illumina mouse-6 v1.0 expression beadchip 3780577 - -
          Illumina mouse-6 v1.0 expression beadchip 5270279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251484 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224437 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224437 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251484 - -
          mouse-6 v1.1 (Illumina) scl0003494.1_23-S - -
          mouse-6 v1.1 (Illumina) scl067905.1_18-S - -
          Illumina Sentrix mouse-6.v1 bead array 3780577 - -
          Illumina Sentrix mouse-6.v1 bead array 5270279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003494.1_23-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067905.1_18-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270279 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3073044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3151840 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251484 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3151840 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3073044 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224437 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251484 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067905.1_18-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline