Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546630_m1
          See other SLC46A1 GE Assays ›
          Gene Symbol:
          Slc46a1
          Gene Name:
          solute carrier family 46, member 1
          Gene Aliases:
          1110002C08Rik, D11Ertd18e, HCP1, Pcft
          Chromosome Location:
          Chr.11: 78465701 - 78471945 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546630_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc46a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026740.2 NP_081016.2 2 - 3 1153 70
          GenBank mRNA AK003278.1 - 2 - 3 1136 70
          AK170505.1 - 2 - 3 1146 70
          AK220201.1 - 2 - 3 1113 70
          BC024522.1 - 1 - 2 289 70
          BC057976.1 - 2 - 3 1138 5052851 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001161.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc46a1

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          folic acid transport
          heme transport
          methotrexate transport
          transmembrane transport
          transporter activity
          folic acid binding
          folic acid transporter activity
          heme transporter activity
          methotrexate transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943492 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943493 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943494 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943495 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943491 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943490 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2168301 BC057976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2168301 NM_026740.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081805 BC057976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081805 AK003278.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081805 BC024522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081805 NM_026740.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647859 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148013 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155518 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137514 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168301 BC057976.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168301 NM_026740.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081805 BC057976.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081805 AK003278.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081805 BC024522.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081805 NM_026740.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430585 AK003278.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27943506 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148013 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155518 - -
          Whole Mouse Genome G4122A A_51_P430585 AK003278.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126374 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122440 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155518 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137514 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148013 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09538191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09557172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09538186 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09538195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09538173 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09538192 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943494 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943490 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943492 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943495 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943493 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27943491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27943482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09584591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09038149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09548839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09621216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09538170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09538192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09538194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09093656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09538191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09245173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09350525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27943474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09557172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09538193 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379187 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379186 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379189 - -
          Affymetrix HT MG-430 PM Array Plate 1426714_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1426715_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379184 - -
          Mouse Genome 430 2.0 Array 1426715_s_at - -
          Mouse Genome 430 2.0 Array 1426714_PM_at - -
          Mouse Genome 430 2.0 Array 1426715_PM_s_at - -
          Mouse Genome 430 2.0 Array 1426714_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52466_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52466_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100010818.mm.1 NM_026740.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010818.mm.1 NM_026740.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010818.mm.1 TC1100001161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010820.mm.1 NM_026740.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010820.mm.1 NM_026740.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010820.mm.1 TC1100001161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010822.mm.1 NM_026740.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010822.mm.1 NM_026740.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010822.mm.1 TC1100001161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010823.mm.1 NM_026740.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010823.mm.1 NM_026740.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010823.mm.1 TC1100001161.mm.1 MTA TC
          Murine Genome U74v2 Set 94339_PM_at - -
          Murine Genome U74v2 Set 94339_at - -
          Affymetrix HT Mouse Genome 430A Array 1426714_at - -
          Affymetrix HT Mouse Genome 430A Array 1426715_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1426715_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1426714_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 418900 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6550300 - -
          Illumina mouse-6 v1.0 expression beadchip 6550300 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2632871 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2632871 - -
          mouse-6 v1.1 (Illumina) scl41202.6.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41202.6.1_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2909275 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2909275 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41202.6.1_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline