Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546635_m1
          See other UCMA GE Assays ›
          Gene Symbol:
          Ucma
          Gene Name:
          upper zone of growth plate and cartilage matrix associated
          Gene Aliases:
          1110017I16Rik, AW121955, Grp
          Chromosome Location:
          Chr.2: 4976122 - 4985748 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546635_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ucma
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113558.2 NP_001107030.1 4 - 5 450 117
          NM_026754.3 NP_081030.1 3 - 4 384 117
          GenBank mRNA AK003750.1 - 3 - 4 326 117
          BC115608.2 - 4 - 5 353 40102702 117
          BC115609.2 - 4 - 5 353 40102703 117
          EF529510.1 - 4 - 5 421 117
          EU368184.1 - 4 - 5 450 117
          FJ217399.1 - 4 - 5 388 117
          FJ217400.1 - 3 - 4 322 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000040.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ucma

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of osteoblast differentiation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 BY590633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 EU368184.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 NM_001113558.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 NM_001165932.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 NM_026754.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 BY590633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 EU368184.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 NM_001113558.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 NM_001165932.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P303217 NM_026754.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609480 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36986 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122881 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 BY590633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 EU368184.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 NM_001113558.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 NM_001165932.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 NM_026754.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 BY590633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 EU368184.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 NM_001113558.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 NM_001165932.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P303217 NM_026754.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20971247 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04414707 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122881 - -
          Whole Mouse Genome G4122A A_51_P303217 BY590633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303217 EU368184.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303217 NM_001113558.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303217 NM_001165932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303217 NM_026754.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303217 BY590633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303217 EU368184.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303217 NM_001113558.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303217 NM_001165932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303217 NM_026754.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139185 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122881 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01582862 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01582861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01582855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20971279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01436038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02263671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01681305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02052382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02052381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01257774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01940990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01257776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01940989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01940994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01157893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01257775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02179964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01304702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01939134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20971281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01940995 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10469064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469061 - -
          Affymetrix HT MG-430 PM Array Plate 1428993_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428994_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10469058 - -
          Mouse Genome 430 2.0 Array 1428994_PM_s_at - -
          Mouse Genome 430 2.0 Array 1428993_at - -
          Mouse Genome 430 2.0 Array 1428993_PM_at - -
          Mouse Genome 430 2.0 Array 1428994_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68527_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68527_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200000324.mm.1 NM_001113558.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000324.mm.1 NM_001113558.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000324.mm.1 NM_001165932.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000324.mm.1 NM_001165932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000324.mm.1 NM_026754.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000324.mm.1 NM_026754.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000324.mm.1 TC0200000040.mm.1 MTA TC
          Mouse Expression Set 430 1428993_PM_at - -
          Mouse Expression Set 430 1428993_at - -
          Mouse Expression Set 430 1428994_s_at - -
          Mouse Expression Set 430 1428994_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1428993_at - -
          Affymetrix HT Mouse Genome 430B Array 1428994_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1428993_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428994_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 550200 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2190082 - -
          Illumina mouse-6 v1.0 expression beadchip 2190082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2702839 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2702839 - -
          mouse-6 v1.1 (Illumina) scl21323.5.1_279-S - -
          Illumina Sentrix mouse-6.v1 bead array 2190082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21323.5.1_279-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2702839 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2702839 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline