Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546738_m1
          See other FAM135B GE Assays ›
          Gene Symbol:
          Fam135b
          Gene Name:
          family with sequence similarity 135, member B
          Gene Aliases:
          1700010C24Rik, A830008O07Rik
          Chromosome Location:
          Chr.15: 71438951 - 71728436 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546738_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam135b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177819.3 NP_808487.2 19 - 20 4195 69
          GenBank mRNA AK005816.1 - 7 - 8 1063 69
          BC119174.1 - 7 - 8 811 69
          BC137579.1 - 7 - 8 811 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001675.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam135b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_51_P156394 BC137579.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156394 AK005816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156394 BC119174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P156394 NM_177819.3 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12321423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12284056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12163085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11942726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12444771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11971847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11919893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11942732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12321436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11884138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12163088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11942742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12122725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11942734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12345131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11942753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12381482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30318419 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10429210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429207 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429211 - -
          Affymetrix Mouse Gene 1.0 ST Array 10429212 - -
          Affymetrix HT MG-430 PM Array Plate 1460477_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443523_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10429203 - -
          Mouse Genome 430 2.0 Array 1443523_at - -
          Mouse Genome 430 2.0 Array 1443523_PM_at - -
          Mouse Genome 430 2.0 Array 1460477_at - -
          Mouse Genome 430 2.0 Array 1460477_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500012567.mm.1 NM_177819.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012567.mm.1 NM_177819.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012567.mm.1 TC1500001675.mm.1 MTA TC
          Mouse Expression Set 430 1460477_PM_at - -
          Mouse Expression Set 430 1443523_PM_at - -
          Mouse Expression Set 430 1443523_at - -
          Mouse Expression Set 430 1460477_at - -
          Affymetrix HT Mouse Genome 430B Array 1460477_at - -
          Affymetrix HT Mouse Genome 430B Array 1443523_at - -
          Affymetrix HT Mouse Genome 430B Array 1460477_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443523_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 359435 - -
          ABI Mouse Genome Survey Microarray 451659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline