Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00547186_m1
          See other SPINK12 GE Assays ›
          Gene Symbol:
          Spink12
          Gene Name:
          serine peptidase inhibitor, Kazal type 12
          Gene Aliases:
          9230117E20Rik
          Chromosome Location:
          Chr.18: 44104523 - 44108543 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00547186_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spink12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030061.3 NP_084337.1 2 - 3 273 73
          GenBank mRNA AK020352.1 - 2 - 3 174 73
          AK136616.1 - 2 - 3 274 73
          BC145914.1 - 2 - 3 159 40131217 73
          BC145916.1 - 2 - 3 159 40131219 73
          DQ437332.1 - 2 - 3 332 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000451.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spink12

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          negative regulation of proteolysis
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955054 AK136616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955054 NM_030061.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296763 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140940 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147544 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955054 AK136616.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955054 NM_030061.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665236 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140940 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147544 - -
          Whole Mouse Genome G4122A A_51_P370552 AK020352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370552 AK136616.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370552 BC145914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370552 BC145916.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370552 DQ437332.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129626 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140940 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147544 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13665930 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13665948 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13665951 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13665936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13665947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13926748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13929326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13934309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14040408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13665934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13948291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13634890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13926749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13868554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14040410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665163 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455448 - -
          Affymetrix HT MG-430 PM Array Plate 1453503_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455446 - -
          Mouse Genome 430 2.0 Array 1453503_PM_at - -
          Mouse Genome 430 2.0 Array 1453503_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78242_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78242_at - -
          Affymetrix Mouse Clariom S Array PSR1800003320.mm.1 NM_030061.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800003320.mm.1 NM_030061.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800003320.mm.1 TC1800000451.mm.1 MTA TC
          Mouse Expression Set 430 1453503_at - -
          Mouse Expression Set 430 1453503_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453503_at - -
          Affymetrix HT Mouse Genome 430B Array 1453503_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 364425 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6370592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2660373 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2660373 - -
          mouse-6 v1.1 (Illumina) scl52011.4.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52011.4.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2660373 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2660373 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0