Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00547547_s1
          See other BLOC1S4 GE Assays ›
          Gene Symbol:
          Bloc1s4
          Gene Name:
          biogenesis of lysosomal organelles complex-1, subunit 4, cappuccino
          Gene Aliases:
          2610101N07Rik, Blos4, Cno
          Chromosome Location:
          Chr.5: 36747374 - 36748679 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00547547_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bloc1s4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133724.3 NP_598485.1 1 763 109
          GenBank mRNA AK011795.1 - 1 735 109
          BC019169.1 - 1 753 5002867 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bloc1s4

          Gene Ontology Categories:

          Function(s) Process(es)

          anterograde axonal transport
          neuron projection development
          melanosome organization
          anterograde synaptic vesicle transport
          neuromuscular process controlling balance
          platelet aggregation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609441 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609438 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609444 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609440 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609439 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098428 BC019169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098428 CK129009.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098428 NM_133724.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098428 AK011795.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678467 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170794 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098428 BC019169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098428 CK129009.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098428 NM_133724.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098428 AK011795.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23609446 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170794 - -
          Whole Mouse Genome G4122A A_51_P390522 BC019169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390522 NM_133724.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P390522 AK011795.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125575 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137415 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170794 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04234862 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04234859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04934104 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609440 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609441 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609438 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23609444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23609432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04472797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04736079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04532301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04518111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04211416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04211418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04569326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04943844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04518101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04313060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04164917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04472799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04736085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04315118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04518100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04890589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04577525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04234858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04530048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04472798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04736080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04504419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04518104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23609378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04530047 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10529580 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529579 - -
          Affymetrix HT MG-430 PM Array Plate 1447620_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1459783_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1418834_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10529577 - -
          Mouse Genome 430 2.0 Array 1418834_PM_at - -
          Mouse Genome 430 2.0 Array 1460160_at - -
          Mouse Genome 430 2.0 Array 1447620_PM_at - -
          Mouse Genome 430 2.0 Array 1447620_at - -
          Mouse Genome 430 2.0 Array 1459783_s_at - -
          Mouse Genome 430 2.0 Array 1459783_PM_s_at - -
          Mouse Genome 430 2.0 Array 1460160_PM_at - -
          Mouse Genome 430 2.0 Array 1418834_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117197_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117197_at - -
          Affymetrix Mouse Clariom S Array PSR0500020857.mm.1 TC0500002263.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020857.mm.1 NM_133724.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020857.mm.1 NM_133724.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020856.mm.1 TC0500002263.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500020856.mm.1 NM_133724.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500020856.mm.1 NM_133724.3 Refseq
          Mouse Expression Set 430 1418834_PM_at - -
          Mouse Expression Set 430 1418834_at - -
          Mouse Expression Set 430 1459783_s_at - -
          Mouse Expression Set 430 1447620_PM_at - -
          Mouse Expression Set 430 1459783_PM_s_at - -
          Mouse Expression Set 430 1447620_at - -
          Murine Genome U74v2 Set 167889_i_at - -
          Murine Genome U74v2 Set 167889_PM_i_at - -
          Murine Genome U74v2 Set 166031_r_at - -
          Murine Genome U74v2 Set 166031_PM_r_at - -
          Murine Genome U74v2 Set 167744_f_at - -
          Murine Genome U74v2 Set 167744_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1418834_PM_at - -
          Mouse Genome 430A 2.0 Array 1418834_at - -
          Affymetrix HT Mouse Genome 430A Array 1418834_at - -
          Affymetrix HT Mouse Genome 430A Array 1418834_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459783_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1447620_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447620_at - -
          Affymetrix HT Mouse Genome 430B Array 1459783_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 489237 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2570048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2591411 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2591411 - -
          mouse-6 v1.1 (Illumina) scl26678.1.8_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26678.1.8_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2591411 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2591411 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl26678.1.8_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline