Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00547967_m1
          See other ALDH16A1 GE Assays ›
          Gene Symbol:
          Aldh16a1
          Gene Name:
          aldehyde dehydrogenase 16 family, member A1
          Gene Aliases:
          2410004H02Rik, AI452234, mFLJ00209
          Chromosome Location:
          Chr.7: 45141833 - 45154538 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00547967_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh16a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145954.1 NP_666066.1 1 - 2 143 89
          GenBank mRNA AK030673.1 - 1 - 2 136 89
          AK144967.1 - 89
          AK144995.1 - 1 - 2 89
          AK151691.1 - 1 - 2 143 89
          AK161198.1 - 1 - 2 181 89
          AK169346.1 - 132 89
          AK170276.1 - 1 - 2 182 89
          AK220200.1 - 1 - 2 119 89
          BC013548.1 - 1 - 2 143 4222058 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002994.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aldh16a1

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          oxidation-reduction process
          aldehyde dehydrogenase (NAD) activity
          oxidoreductase activity
          oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25091454 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122362 BC013548.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122362 BC030467.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122362 AK170276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122362 AK220200.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122362 AK161198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017755 AK164374.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714168 AK079352.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81326 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017755 AK164374.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122362 BC013548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122362 BC030467.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122362 AK170276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122362 AK220200.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122362 AK161198.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091480 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01507238 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01507222 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06182071 - -
          Whole Mouse Genome G4122A A_51_P407311 BC013548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407311 BC030467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407311 AK170276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407311 AK220200.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407311 AK161198.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407304 BC030467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407304 AK220200.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407304 AK169346.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407304 AK151691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407304 AK030673.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122362 BC013548.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122362 BC030467.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122362 AK170276.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122362 AK220200.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122362 AK161198.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109850 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01507238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06258860 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06258858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06258856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06258861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05885451 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05885454 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25091454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06371931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06430758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06419571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06371937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06180578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06258860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06419584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06430766 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563135 - -
          Affymetrix HT MG-430 PM Array Plate 1447372_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423731_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563130 - -
          Mouse Genome 430 2.0 Array 1447372_PM_at - -
          Mouse Genome 430 2.0 Array 1447372_at - -
          Mouse Genome 430 2.0 Array 1423731_at - -
          Mouse Genome 430 2.0 Array 1423731_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69748_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69748_at - -
          Affymetrix Mouse Clariom S Array PSR0700026990.mm.1 NM_145954.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026990.mm.1 NM_145954.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026990.mm.1 TC0700002994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026993.mm.1 NM_145954.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026993.mm.1 NM_145954.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026993.mm.1 TC0700002994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026991.mm.1 NM_145954.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026991.mm.1 NM_145954.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026991.mm.1 TC0700002994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026992.mm.1 NM_145954.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026992.mm.1 NM_145954.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026992.mm.1 TC0700002994.mm.1 MTA TC
          Mouse Expression Set 430 1447372_at - -
          Mouse Expression Set 430 1447372_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423731_at - -
          Affymetrix HT Mouse Genome 430A Array 1423731_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447372_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447372_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913029 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5050373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050373 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245112 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245112 - -
          mouse-6 v1.1 (Illumina) scl31390.16.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050373 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31390.16.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050373 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245112 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245112 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline