Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00549254_m1
          See other MYL6B GE Assays ›
          Gene Symbol:
          Myl6b
          Gene Name:
          myosin, light polypeptide 6B
          Gene Aliases:
          5730437E04Rik, BC037527, MLC1SA
          Chromosome Location:
          Chr.10: 128494157 - 128498685 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00549254_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myl6b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172259.1 NP_758463.1 2 - 3 275 81
          GenBank mRNA BC037527.1 - 2 - 3 275 3468485 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003146.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myl6b

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          skeletal muscle tissue development
          muscle filament sliding
          motor activity
          calcium ion binding
          structural constituent of muscle

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952990 NM_172259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952990 BC037527.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176611 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110466 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952990 NM_172259.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952990 BC037527.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541708 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110466 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02247437 - -
          Whole Mouse Genome G4122A A_52_P578692 NM_172259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P578692 BC037527.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476725 AK017625.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112319 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116488 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110466 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08475750 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08719260 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08591012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08591009 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08475748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08475745 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08719263 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08719261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08591010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08827920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08498003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08283091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08827917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08827907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08827916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08636057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08827902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08929837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08878136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541721 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373402 - -
          Affymetrix HT MG-430 PM Array Plate 1430661_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373400 - -
          Mouse Genome 430 2.0 Array 1430661_PM_at - -
          Mouse Genome 430 2.0 Array 1430661_at - -
          Affymetrix Mouse Clariom S Array PSR1000023612.mm.1 NM_172259.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023612.mm.1 NM_172259.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023612.mm.1 TC1000003146.mm.1 MTA TC
          Mouse Expression Set 430 1430661_PM_at - -
          Mouse Expression Set 430 1430661_at - -
          Murine Genome U74v2 Set 92670_PM_at - -
          Murine Genome U74v2 Set 92670_at - -
          Murine Genome U74v2 Set 97101_at - -
          Murine Genome U74v2 Set 97101_PM_at - -
          Mouse Genome 430A 2.0 Array 1430661_at - -
          Mouse Genome 430A 2.0 Array 1430661_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1430661_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1430661_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 468349 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 101940452 - -
          Illumina mouse-6 v1.0 expression beadchip 6400546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006400546 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737183 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240281 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737183 - -
          mouse-6 v1.1 (Illumina) scl38279.2_20-S - -
          mouse-6 v1.1 (Illumina) scl37360.7.1_32-S - -
          Illumina Sentrix mouse-6.v1 bead array 101940452 - -
          Illumina Sentrix mouse-6.v1 bead array 6400546 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37360.7.1_32-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101940452 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6400546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2737183 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240281 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2737183 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37360.7.1_32-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline