Search
Search

Mark4| Gene Symbol: Mark4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172279.1 | NP_758483.1 | 13 - 14 | 1496 | 63 | |
| GenBank mRNA | AK035668.1 | - | 1 - 2 | 164 | 63 | |
| AK122565.2 | - | 9 - 10 | 1142 | 63 | ||
| AK146784.1 | - | 14 - 15 | 1831 | 63 | ||
| AK207936.1 | - | 1 - 2 | 146 | 63 | ||
| AK216688.1 | - | 1 - 2 | 140 | 63 | ||
| AY151083.1 | - | 13 - 14 | 1496 | 63 | ||
Set Membership: |
|