Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00549528_m1
          See other ZFP759 GE Assays ›
          Gene Symbol:
          Zfp759
          Gene Name:
          zinc finger protein 759
          Gene Aliases:
          BC028265, Rslcan-8, Rslcan8
          Chromosome Location:
          Chr.13: 67125128 - 67142404 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00549528_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp759
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172392.3 NP_765980.3 3 - 4 306 101
          GenBank mRNA AK030126.1 - 3 - 4 309 101
          BC028265.1 - 3 - 4 290 5365910 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000817.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC028265.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp759

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P491554 BC028265.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P491554 NM_172392.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P491554 AK030126.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P491554 BC028265.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P491554 NM_172392.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P491554 AK030126.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313216 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12666 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360346 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 100654 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176591 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174779 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121813 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P491554 BC028265.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P491554 NM_172392.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P491554 AK030126.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P491554 BC028265.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P491554 NM_172392.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P491554 AK030126.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29107497 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07905175 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121813 - -
          Whole Mouse Genome G4122A A_52_P491554 BC028265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P491554 NM_172392.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P491554 AK030126.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P491554 BC028265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P491554 NM_172392.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P491554 AK030126.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124293 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113756 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121813 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10596095 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11113488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10596104 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11113492 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10596090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29107487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11103300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10756967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10490821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11113491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29107474 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405914 - -
          Affymetrix HT MG-430 PM Array Plate 1452623_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1445270_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10405911 - -
          Mouse Genome 430 2.0 Array 1445270_at - -
          Mouse Genome 430 2.0 Array 1445270_PM_at - -
          Mouse Genome 430 2.0 Array 1452623_at - -
          Mouse Genome 430 2.0 Array 1452623_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 268670_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 268670_at - -
          Affymetrix Mouse Clariom S Array PSR1300005339.mm.1 NM_172392.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005339.mm.1 NM_172392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005339.mm.1 TC1300000817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005341.mm.1 NM_172392.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005341.mm.1 NM_172392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005341.mm.1 TC1300000817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005340.mm.1 NM_172392.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005340.mm.1 NM_172392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005340.mm.1 TC1300000817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005342.mm.1 NM_172392.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005342.mm.1 NM_172392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005342.mm.1 TC1300000817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005343.mm.1 NM_172392.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005343.mm.1 NM_172392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005343.mm.1 TC1300000817.mm.1 MTA TC
          Mouse Expression Set 430 1452623_at - -
          Mouse Expression Set 430 1452623_PM_at - -
          Mouse Genome 430A 2.0 Array 1452623_at - -
          Mouse Genome 430A 2.0 Array 1452623_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452623_at - -
          Affymetrix HT Mouse Genome 430A Array 1452623_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 757403 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2940341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256222 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256222 - -
          mouse-6 v1.1 (Illumina) scl44666.7_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44666.7_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256222 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256222 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl44666.7_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline