Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00549650_m1
          See other GPC2 GE Assays ›
          Gene Symbol:
          Gpc2
          Gene Name:
          glypican 2 (cerebroglycan)
          Gene Aliases:
          2410016G05Rik
          Chromosome Location:
          Chr.5: 138273657 - 138279958 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00549650_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172412.2 NP_766000.1 3 - 4 772 112
          GenBank mRNA AK010515.1 - 3 - 4 802 112
          AK048605.1 - 3 - 4 715 112
          AK049639.1 - 3 - 4 773 112
          AK160959.1 - 3 - 4 818 112
          BC056941.1 - 2 - 3 1471 112
          BC083180.1 - 3 - 4 779 5707617 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003741.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpc2

          Gene Ontology Categories:

          Function(s) Process(es)

          smoothened signaling pathway
          neuron differentiation
          heparan sulfate proteoglycan binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129656 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129622 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129654 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129620 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129652 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 AK160959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 BC056941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 BC083180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 AK048605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 AK049639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 AK160959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 BC056941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 BC083180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 AK048605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185627 AK049639.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839725 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144339 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 AK160959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 BC056941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 BC083180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 AK048605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 AK049639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 AK160959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 BC056941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 BC083180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 AK048605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185627 AK049639.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129657 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05762629 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144339 - -
          Whole Mouse Genome G4122A A_51_P185627 AK160959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185627 BC056941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185627 BC083180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185627 AK048605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185627 AK049639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185627 AK160959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185627 BC056941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185627 BC083180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185627 AK048605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185627 AK049639.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131496 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144339 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04874432 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04874448 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04874443 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04874446 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04874444 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129656 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129620 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129622 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129651 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129652 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129655 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129654 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04874435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04874438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04874445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04874431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04874440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04951613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04874437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04951614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04645687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10535022 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535023 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535024 - -
          Affymetrix HT MG-430 PM Array Plate 1429293_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10535021 - -
          Mouse Genome 430 2.0 Array 1429293_PM_at - -
          Mouse Genome 430 2.0 Array 1429293_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71951_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71951_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500031785.mm.1 TC0500003741.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031785.mm.1 NM_172412.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031785.mm.1 NM_172412.2 Refseq
          Mouse Expression Set 430 1429293_PM_at - -
          Mouse Expression Set 430 1429293_at - -
          Murine Genome U74v2 Set 166160_PM_f_at - -
          Murine Genome U74v2 Set 166160_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1429293_at - -
          Affymetrix HT Mouse Genome 430B Array 1429293_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 906895 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 130647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000130647 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2696721 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2696721 - -
          mouse-6 v1.1 (Illumina) scl25856.3.1_318-S - -
          Illumina Sentrix mouse-6.v1 bead array 130647 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25856.3.1_318-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 130647 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2696721 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2696721 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0