Search
Search

Arhgef10l| Gene Symbol: Arhgef10l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112722.1 | NP_001106193.1 | 26 - 27 | 3249 | 54 | |
| NM_001112723.1 | NP_001106194.1 | 25 - 26 | 3207 | 54 | ||
| NM_001290803.1 | NP_001277732.1 | 27 - 28 | 3348 | 54 | ||
| NM_172415.3 | NP_766003.3 | 26 - 27 | 3324 | 54 | ||
| GenBank mRNA | AK028570.1 | - | 20 - 21 | 2665 | 54 | |
| AK028648.1 | - | 26 - 27 | 3232 | 54 | ||
| AK129408.1 | - | 25 - 26 | 3207 | 54 | ||
| BC021843.1 | - | 12 - 13 | 1494 | 54 | ||
| BC024646.1 | - | 1 - 2 | 239 | 54 | ||
| BC034061.1 | - | 20 - 21 | 2485 | 54 | ||
| BC059220.1 | - | 26 - 27 | 3312 | 6417641 | 54 | |
Set Membership: |
|