Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00549967_m1
          See other SLC38A5 GE Assays ›
          Gene Symbol:
          Slc38a5
          Gene Name:
          solute carrier family 38, member 5
          Gene Aliases:
          C81234, E330031E14, JM24, SN2
          Chromosome Location:
          Chr.X: 8271381 - 8280179 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00549967_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc38a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172479.3 NP_766067.2 7 - 8 618 73
          GenBank mRNA AK054485.1 - 7 - 8 605 73
          AK077667.1 - 7 - 8 587 73
          AK155928.1 - 7 - 8 571 73
          AK158125.1 - 6 - 7 739 73
          BC152401.1 - 7 - 8 587 40057282 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000088.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc38a5

          Panther Classification:

          Molecular Function -

          amino acid transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          neutral amino acid transport
          glycine transport
          cation transmembrane transport
          L-alpha-amino acid transmembrane transport
          amino acid transmembrane transporter activity
          neutral amino acid transmembrane transporter activity
          glycine transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010641 BX523289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010641 NM_172479.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335337 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565309 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 523328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 708452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328417 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114300 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010641 BX523289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010641 NM_172479.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400269 BX523289.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400269 AK155928.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400269 BC058856.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400269 NM_172479.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400269 AK054485.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32242518 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114300 - -
          Whole Mouse Genome G4122A A_51_P400269 BX523289.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400269 AK155928.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400269 BC058856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400269 NM_172479.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400269 AK054485.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132590 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114300 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15107089 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15107087 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15107092 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15107090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32242487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15192226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15192224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15160588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15122105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15122119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15107092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14708403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32242576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15192211 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598517 - -
          Affymetrix HT MG-430 PM Array Plate 1454622_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10598507 - -
          Mouse Genome 430 2.0 Array 1454622_PM_at - -
          Mouse Genome 430 2.0 Array 1454622_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 209837_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 209837_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00000669.mm.1 NM_172479.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000669.mm.1 TC0X00000088.mm.1 MTA TC
          Mouse Expression Set 430 1454622_at - -
          Mouse Expression Set 430 1454622_PM_at - -
          Murine Genome U74v2 Set 160907_at - -
          Murine Genome U74v2 Set 160907_PM_at - -
          Mouse Genome 430A 2.0 Array 1454622_PM_at - -
          Mouse Genome 430A 2.0 Array 1454622_at - -
          Affymetrix HT Mouse Genome 430A Array 1454622_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1454622_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 652756 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 540093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2649773 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2649773 - -
          mouse-6 v1.1 (Illumina) scl55061.19.1_283-S - -
          Illumina Sentrix mouse-6.v1 bead array 540093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl55061.19.1_283-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540093 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2649773 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2649773 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl55061.19.1_283-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline