Search
Search
| Gene Symbol: Araf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159645.1 | NP_001153117.1 | 2 - 3 | 234 | 80 | |
| NM_009703.2 | NP_033833.1 | 2 - 3 | 234 | 80 | ||
| GenBank mRNA | AB158252.1 | - | 1 - 2 | 159 | 80 | |
| AK004741.1 | - | 1 - 2 | 1513 | 80 | ||
| AK020547.1 | - | 2 - 3 | 200 | 80 | ||
| AK034861.1 | - | 2 - 3 | 229 | 80 | ||
| AK039044.1 | - | 2 - 3 | 200 | 80 | ||
| AK085823.1 | - | 2 - 3 | 200 | 80 | ||
| AK133797.1 | - | 1 - 2 | 1520 | 80 | ||
| AK135358.1 | - | 2 - 3 | 200 | 80 | ||
| AK163567.1 | - | 2 - 3 | 203 | 80 | ||
| AK169757.1 | - | 2 - 3 | 215 | 80 | ||
| BB857915.1 | - | 2 - 3 | 234 | 80 | ||
| BC004757.1 | - | 2 - 3 | 190 | 3154293 | 80 | |
Set Membership: |
|