Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00550786_m1
          See other CCDC163 GE Assays ›
          Gene Symbol:
          Ccdc163
          Gene Name:
          coiled-coil domain containing 163
          Gene Aliases:
          0610037D15Rik, 4933430J04Rik
          Chromosome Location:
          Chr.4: 116708555 - 116715104 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00550786_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc163
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026714.2 NP_080990.2 8 - 9 890 72
          GenBank mRNA AI606993.1 - 4 - 5 422 72
          AK002768.1 - 8 - 9 792 72
          AW121394.1 - 2 - 3 72
          BC022718.1 - 7 - 8 889 72
          BC022733.1 - 8 - 9 913 4506628 72
          CD741104.1 - 2 - 3 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004189.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc163

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144636 BC022733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144636 CD741104.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144636 NM_026714.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2144636 BC022718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2269700 BC022733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2269700 BG297132.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2269700 DT911344.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2269700 NM_026714.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2269700 AK016995.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164720 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177885 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2269700 BC022733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2269700 BG297132.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2269700 DT911344.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2269700 NM_026714.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2269700 AK016995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144636 BC022733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144636 CD741104.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144636 NM_026714.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2144636 BC022718.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23230583 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177885 - -
          Whole Mouse Genome G4122A A_51_P321405 BC022733.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321405 BC022718.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115698 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116554 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03329405 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03879487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23230596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03674593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03329397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03674591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03329407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23230597 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10507346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507342 - -
          Affymetrix HT MG-430 PM Array Plate 1453639_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1425043_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10507334 - -
          Mouse Genome 430 2.0 Array 1453639_PM_s_at - -
          Mouse Genome 430 2.0 Array 1425043_s_at - -
          Mouse Genome 430 2.0 Array 1453639_s_at - -
          Mouse Genome 430 2.0 Array 1425043_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68394_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68394_at - -
          Affymetrix Mouse Clariom S Array PSR0400009425.mm.1 NM_026714.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400009425.mm.1 NM_026714.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400009425.mm.1 TC0400004189.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400009423.mm.1 NM_026714.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400009423.mm.1 NM_026714.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400009423.mm.1 TC0400004189.mm.1 MTA TC
          Mouse Expression Set 430 1425043_s_at - -
          Mouse Expression Set 430 1425043_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1425043_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1425043_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1425043_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1425043_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1453639_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1453639_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 747173 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6840463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234456 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234456 - -
          mouse-6 v1.1 (Illumina) scl068394.8_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068394.8_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234456 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234456 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline