| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27534909 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27534907 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27534906 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27534908 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
BC004613.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
AK144464.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
AK144505.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
AK164021.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
NM_001190454.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
BC004613.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
AK144464.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
AK144505.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
AK164021.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P435333 |
NM_001190454.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
783867 |
AK052703.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
783867 |
AK144030.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
783867 |
NM_172532.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
822642 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
187586 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
410822 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
595881 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674189 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731078 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
153309 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
250342 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
653650 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
151019 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
89836 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P111108 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P161958 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
BC004613.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
AK144464.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
AK144505.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
AK164021.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
NM_001190454.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
BC004613.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
AK144464.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
AK144505.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
AK164021.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P435333 |
NM_001190454.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
BC004613.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
AK144464.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
AK144505.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
AK164021.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
NM_001190454.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
BC004613.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
AK144464.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
AK144505.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
AK164021.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P435333 |
NM_001190454.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534920 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534916 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534909 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534907 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534924 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534914 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534919 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534925 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534908 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534913 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534911 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534906 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27534917 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02245375 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07229802 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P111108 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P161958 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P435333 |
BC004613.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435333 |
AK144464.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435333 |
AK144505.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435333 |
AK164021.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435333 |
NM_001190454.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435333 |
BC004613.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435333 |
AK144464.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435333 |
AK144505.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435333 |
AK164021.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P435333 |
NM_001190454.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P106250 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P161958 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P111108 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08263377 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27534909 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27534907 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27534908 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27534906 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535051 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535064 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534946 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535085 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535057 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535083 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535039 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534974 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535047 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534913 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535081 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535036 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535044 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534956 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535078 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535030 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534906 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534942 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534926 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535007 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534900 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534887 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534909 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534937 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534896 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535053 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534954 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534892 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534902 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534879 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535084 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534916 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534881 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535016 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534977 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534895 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535067 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534852 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535004 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534878 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534985 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535028 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534855 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535049 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534957 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534862 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534889 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534873 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534869 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534884 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535035 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534914 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534875 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534996 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534983 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534905 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535027 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534911 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535046 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535069 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534970 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534903 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534871 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535015 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534931 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534932 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535022 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534989 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535066 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535040 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535055 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535059 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534967 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535025 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534943 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535013 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534945 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534924 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535038 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535080 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534972 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535090 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534928 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534865 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534949 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534993 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534939 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534898 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534925 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534990 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535087 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534859 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535042 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535074 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535061 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27535009 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534968 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534919 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27534874 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534932 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534968 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08684266 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534967 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534898 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534900 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08669630 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08669638 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534974 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27535009 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27535000 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534908 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534996 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08263381 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534935 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534946 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534954 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534911 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534896 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534986 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08263369 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27535007 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534909 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08492004 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534925 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27535001 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534906 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08492011 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534917 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534919 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534924 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08933857 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534913 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534991 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534949 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08491994 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27535004 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534983 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534903 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534964 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534982 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534918 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534960 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08595158 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534945 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27535002 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534920 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534957 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534941 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534999 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534993 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534916 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534931 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08669643 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08684255 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534985 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534937 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534962 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534939 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08684254 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08684268 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08491996 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534928 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08684249 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08933871 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27534914 |
- |
- |