Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00550901_g1
          See other PPP1R35 GE Assays ›
          Gene Symbol:
          Ppp1r35
          Gene Name:
          protein phosphatase 1, regulatory subunit 35
          Gene Aliases:
          2010007H12Rik, 2010011D20Rik, AL024004
          Chromosome Location:
          Chr.5: 137778904 - 137782201 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00550901_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r35
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027242.4 NP_081518.1 3 - 4 687 97
          GenBank mRNA AK008148.1 - 3 - 4 688 97
          AK008184.1 - 3 - 4 684 97
          BC052497.1 - 3 - 4 663 1429227 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001635.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r35

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of phosphatase activity
          protein phosphatase inhibitor activity
          phosphatase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127509 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103865 AK008184.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103865 AK008148.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103865 AK008184.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103865 AK008148.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103865 AK008184.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103865 AK008148.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103865 AK008184.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103865 AK008148.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24127493 - -
          Whole Mouse Genome G4122A A_51_P103865 AK008184.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103865 AK008148.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103865 AK008184.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103865 AK008148.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04616952 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04616945 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04546183 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24127507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24127580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04616945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04807352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04683022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04354646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04613610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04759585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04301091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04733273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04104211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24127486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04350108 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526691 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526690 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526692 - -
          Affymetrix HT MG-430 PM Array Plate 1429255_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526687 - -
          Mouse Genome 430 2.0 Array 1429255_PM_at - -
          Mouse Genome 430 2.0 Array 1429255_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69871_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69871_at - -
          Affymetrix Mouse Clariom S Array PSR0500014935.mm.1 TC0500001635.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014935.mm.1 NM_027242.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014935.mm.1 NM_027242.4 Refseq
          Affymetrix HT Mouse Genome 430B Array 1429255_at - -
          Affymetrix HT Mouse Genome 430B Array 1429255_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 634871 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1230270 - -
          MouseWG-6 v2 Expression BeadChip 7050162 - -
          Illumina mouse-6 v1.0 expression beadchip 7050162 - -
          Illumina mouse-6 v1.0 expression beadchip 1230270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230270 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2662283 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227103 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227103 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2662283 - -
          mouse-6 v1.1 (Illumina) scl27092.3.1_116-S - -
          mouse-6 v1.1 (Illumina) scl0004036.1_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 7050162 - -
          Illumina Sentrix mouse-6.v1 bead array 1230270 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0004036.1_27-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27092.3.1_116-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230270 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2662283 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227103 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2662283 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227103 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27092.3.1_116-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0004036.1_27-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline