Search
Search

Ankrd42| Gene Symbol: Ankrd42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028665.4 | NP_082941.2 | 4 - 5 | 1409 | 90 | |
| GenBank mRNA | AK029511.1 | - | 4 - 5 | 1411 | 90 | |
| AK029888.1 | - | 4 - 5 | 1370 | 90 | ||
| AK033599.1 | - | 4 - 5 | 1398 | 90 | ||
| AK048925.1 | - | 4 - 5 | 1303 | 90 | ||
| AK133339.1 | - | 1370 | 90 | |||
| BC030885.1 | - | 4 - 5 | 1038 | 90 | ||
| BC145456.1 | - | 4 - 5 | 527 | 9054017 | 90 | |
Set Membership: |
|