Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00551746_m1
          See other SLC34A3 GE Assays ›
          Gene Symbol:
          Slc34a3
          Gene Name:
          solute carrier family 34 (sodium phosphate), member 3
          Gene Aliases:
          AI649385, Npt2c, Npt2c-v1, NptIIc, naPi-2c
          Chromosome Location:
          Chr.2: 25228897 - 25234271 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00551746_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc34a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080854.3 NP_543130.2 12 - 13 1432 64
          GenBank mRNA AB054999.1 - 12 - 13 1412 64
          BC125326.1 - 11 - 12 1365 40129841 64
          BC131997.1 - 11 - 12 1365 40130274 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003137.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc34a3

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          sodium ion transport
          phosphate ion transport
          cellular phosphate ion homeostasis
          phosphate ion transmembrane transport
          sodium ion transmembrane transport
          sodium-dependent phosphate transport
          sodium:phosphate symporter activity
          symporter activity
          sodium-dependent phosphate transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230734 BX637671.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230734 NM_080854.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230734 AB054999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230734 BX637671.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230734 NM_080854.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230734 AB054999.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736055 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116461 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230734 BX637671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230734 NM_080854.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230734 AB054999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230734 BX637671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230734 NM_080854.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230734 AB054999.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230734 BX637671.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230734 NM_080854.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230734 AB054999.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230734 BX637671.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230734 NM_080854.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230734 AB054999.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070728 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00324627 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116461 - -
          Whole Mouse Genome G4122A A_51_P230734 BX637671.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230734 NM_080854.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230734 AB054999.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230734 BX637671.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230734 NM_080854.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230734 AB054999.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104648 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101813 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01295977 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01296040 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01296020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01295981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01296053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01296022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01295978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070730 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10480641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480636 - -
          Affymetrix HT MG-430 PM Array Plate 1439519_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10480633 - -
          Mouse Genome 430 2.0 Array 1439519_PM_at - -
          Mouse Genome 430 2.0 Array 1439519_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 142681_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 142681_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200025768.mm.1 NM_080854.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025768.mm.1 NM_080854.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025768.mm.1 TC0200003137.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025767.mm.1 NM_080854.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025767.mm.1 NM_080854.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025767.mm.1 TC0200003137.mm.1 MTA TC
          Mouse Expression Set 430 1439519_PM_at - -
          Mouse Expression Set 430 1439519_at - -
          Affymetrix HT Mouse Genome 430B Array 1439519_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439519_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 613304 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2230242 - -
          Illumina mouse-6 v1.0 expression beadchip 2230242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258111 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258111 - -
          mouse-6 v1.1 (Illumina) scl19575.14.1_212-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19575.14.1_212-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258111 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258111 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline