Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00551904_s1
          See other OLFR993 GE Assays ›
          Gene Symbol:
          Olfr993
          Gene Name:
          olfactory receptor 993
          Gene Aliases:
          MOR203-2
          Chromosome Location:
          Chr.2: 85413933 - 85414877 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00551904_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr993
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146435.1 NP_666646.1 1 353 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003894.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr993

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967573 NM_146435.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149529 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967573 NM_146435.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P418682 NM_146435.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149529 - -
          Whole Mouse Genome G4122A A_51_P418682 NM_146435.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110183 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01187465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01863429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397816 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484515 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484514 - -
          Affymetrix Mouse Clariom S Array PSR0200033685.mm.1 NM_146435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033685.mm.1 NM_146435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033685.mm.1 TC0200003894.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5050273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1250193 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250193 - -
          mouse-6 v1.1 (Illumina) scl4361.1.1_31-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050273 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4361.1.1_31-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050273 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1250193 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1250193 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0