Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00553091_m1
          See other GNPTG GE Assays ›
          Gene Symbol:
          Gnptg
          Gene Name:
          N-acetylglucosamine-1-phosphotransferase, gamma subunit
          Gene Aliases:
          6430527N14Rik, A830081F19, AU067667, AU067744, Mdcp1, Tce7
          Chromosome Location:
          Chr.17: 25234317 - 25240170 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00553091_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnptg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172529.3 NP_766117.2 3 - 4 183 66
          GenBank mRNA AK044021.1 - 2 - 3 263 66
          AK078230.1 - 191 66
          AY465529.1 - 3 - 4 191 66
          BC023869.1 - 2 - 3 266 66
          BC055872.1 - 3 - 4 183 4925332 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001655.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnptg

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          N-glycan processing to lysosome
          carbohydrate phosphorylation
          UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosaminephosphotransferase activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102480 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102481 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124543 BC055872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124543 AK078230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124543 NM_172529.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124543 BC089378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124543 BC059910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011087 BC023869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011087 BC055872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011087 AK078230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011087 AK044021.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011087 NM_172529.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891640 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290999 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102846 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104983 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138148 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124543 BC055872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124543 AK078230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124543 NM_172529.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124543 BC089378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124543 BC059910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011087 BC023869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011087 BC055872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011087 AK078230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011087 AK044021.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011087 NM_172529.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491017 AK078230.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491017 BC059910.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491017 NM_028789.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491017 AK004898.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491017 NM_001197024.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102462 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104983 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138148 - -
          Whole Mouse Genome G4122A A_51_P491017 AK078230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491017 BC059910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491017 NM_028789.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491017 AK004898.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491017 NM_001197024.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665928 AK078230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665928 AK044021.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665928 NM_172529.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102306 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104983 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138148 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13506769 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13506758 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13175531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102481 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13552278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13485417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13552277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13175531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13175532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13485372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13485362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13485421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13485381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13073941 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448870 - -
          Affymetrix HT MG-430 PM Array Plate 1436526_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448865 - -
          Mouse Genome 430 2.0 Array 1436526_at - -
          Mouse Genome 430 2.0 Array 1436526_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214505_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214505_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700014989.mm.1 NM_172529.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014989.mm.1 NM_172529.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014989.mm.1 TC1700001655.mm.1 MTA TC
          Mouse Expression Set 430 1436526_at - -
          Mouse Expression Set 430 1436526_PM_at - -
          Murine Genome U74v2 Set 98621_at - -
          Murine Genome U74v2 Set 98621_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436526_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436526_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 418927 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1740546 - -
          Illumina mouse-6 v1.0 expression beadchip 1740546 - -
          Illumina mouse-6 v1.0 expression beadchip 2030273 - -
          Illumina mouse-6 v1.0 expression beadchip 3140176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003140176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731999 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2617865 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731999 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2617865 - -
          mouse-6 v1.1 (Illumina) scl0214505.1_41-S - -
          mouse-6 v1.1 (Illumina) scl00214505.2_7-S - -
          mouse-6 v1.1 (Illumina) scl00214505.2_157-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030273 - -
          Illumina Sentrix mouse-6.v1 bead array 1740546 - -
          Illumina Sentrix mouse-6.v1 bead array 3140176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0214505.1_41-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00214505.2_7-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00214505.2_157-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3140176 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740546 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030273 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2617865 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2731999 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226861 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2617865 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2731999 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226861 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00214505.2_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0214505.1_41-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline