Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00553269_m1
          See other YBEY GE Assays ›
          Gene Symbol:
          Ybey
          Gene Name:
          ybeY metallopeptidase
          Gene Aliases:
          1110068N23Rik, A130042E20Rik, C21orf57, ORF57, ORF66
          Chromosome Location:
          Chr.10: 76459567 - 76469114 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00553269_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ybey
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172550.4 NP_766138.3 3 - 4 700 73
          GenBank mRNA AK037727.1 - 3 - 4 427 73
          AK136543.1 - 3 - 4 451 73
          BC057308.1 - 3 - 4 700 5703033 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002401.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ybey

          Panther Classification:

          Molecular Function -

          RNA metabolism protein endoribonuclease nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          biological_process
          metalloendopeptidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27262216 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459091 AK149490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459091 BC057308.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459091 AK037727.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459091 NM_172550.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459091 AK149490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459091 BC057308.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459091 AK037727.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P459091 NM_172550.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 846581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42395 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142383 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459091 AK149490.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459091 BC057308.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459091 AK037727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459091 NM_172550.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459091 AK149490.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459091 BC057308.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459091 AK037727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P459091 NM_172550.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27262244 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02163061 - -
          Whole Mouse Genome G4122A A_51_P459091 AK149490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459091 BC057308.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459091 AK037727.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459091 NM_172550.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459091 AK149490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459091 BC057308.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459091 AK037727.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P459091 NM_172550.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113422 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08891917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08302226 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08891905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27262216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27262263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08302228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08891923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08891908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08891915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27262235 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10370179 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364163 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370177 - -
          Affymetrix HT MG-430 PM Array Plate 1435100_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10364155 - -
          Affymetrix Mouse Gene 1.1 ST Array 10370174 - -
          Mouse Genome 430 2.0 Array 1435100_at - -
          Mouse Genome 430 2.0 Array 1435100_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 216119_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 216119_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000017321.mm.1 NM_172550.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017321.mm.1 NM_172550.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017321.mm.1 TC1000002401.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017322.mm.1 NM_172550.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017322.mm.1 NM_172550.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017322.mm.1 TC1000002401.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430B Array 1435100_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435100_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 531356 - -
          ABI Mouse Genome Survey Microarray 566875 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6370048 - -
          Illumina mouse-6 v1.0 expression beadchip 6370048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220132 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673460 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220132 - -
          mouse-6 v1.1 (Illumina) scl0216119.1_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0216119.1_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220132 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220132 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline