Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00553764_m1
          See other PHF11A GE Assays ›
          Gene Symbol:
          Phf11a
          Gene Name:
          PHD finger protein 11A
          Gene Aliases:
          4933417L10Rik, Phf11, Phf11l
          Chromosome Location:
          Chr.14: 59276913 - 59297522 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00553764_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phf11a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172603.3 NP_766191.1 7 - 8 692 78
          GenBank mRNA AK077176.1 - 7 - 8 687 78
          BC116758.1 - 7 - 8 676 40126077 78
          BC116760.1 - 7 - 8 676 40126079 78
          EU155109.1 - 7 - 8 676 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002161.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phf11a

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of immune response
          molecular_function
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262515 EU155109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262515 NM_172603.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262515 AK077176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262515 EU155109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262515 NM_172603.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262515 AK077176.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13444 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262515 EU155109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262515 NM_172603.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262515 AK077176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262515 EU155109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262515 NM_172603.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262515 AK077176.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08127549 - -
          Whole Mouse Genome G4122A A_51_P262515 EU155109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262515 NM_172603.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262515 AK077176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262515 EU155109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262515 NM_172603.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262515 AK077176.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116728 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134493 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08127549 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11702505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29637775 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10420485 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420484 - -
          Affymetrix Mouse Gene 1.1 ST Array 10420483 - -
          Affymetrix Mouse Clariom S Array PSR1400016670.mm.1 NM_172603.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016670.mm.1 NM_172603.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016670.mm.1 TC1400002161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016665.mm.1 NM_172603.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016665.mm.1 NM_172603.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016665.mm.1 TC1400002161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016666.mm.1 NM_172603.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016666.mm.1 NM_172603.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016666.mm.1 TC1400002161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016668.mm.1 NM_172603.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016668.mm.1 NM_172603.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016668.mm.1 TC1400002161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016669.mm.1 NM_172603.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016669.mm.1 NM_172603.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016669.mm.1 TC1400002161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016663.mm.1 NM_172603.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016663.mm.1 NM_172603.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016663.mm.1 TC1400002161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016671.mm.1 NM_172603.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016671.mm.1 NM_172603.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016671.mm.1 TC1400002161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016664.mm.1 NM_172603.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016664.mm.1 NM_172603.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016664.mm.1 TC1400002161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016667.mm.1 NM_172603.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016667.mm.1 NM_172603.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016667.mm.1 TC1400002161.mm.1 MTA TC
          Murine Genome U74v2 Set 135384_PM_at - -
          Murine Genome U74v2 Set 135384_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 667991 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1170358 - -
          MouseWG-6 v2 Expression BeadChip 630025 - -
          Illumina mouse-6 v1.0 expression beadchip 1170358 - -
          Illumina mouse-6 v1.0 expression beadchip 630025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170358 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2696492 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225451 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2696492 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225451 - -
          mouse-6 v1.1 (Illumina) scl0219131.1_319-S - -
          mouse-6 v1.1 (Illumina) scl00219131.1_101-S - -
          Illumina Sentrix mouse-6.v1 bead array 630025 - -
          Illumina Sentrix mouse-6.v1 bead array 1170358 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0219131.1_319-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00219131.1_101-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630025 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170358 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2696492 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2696491 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225451 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2696492 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2696491 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0