Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00553947_m1
          See other TREML4 GE Assays ›
          Gene Symbol:
          Treml4
          Gene Name:
          triggering receptor expressed on myeloid cells-like 4
          Gene Aliases:
          5031403H21Rik, BB137214, IDCP1, TLT-4, TLT4, Treml3
          Chromosome Location:
          Chr.17: 48264191 - 48275607 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00553947_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Treml4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033922.2 NP_001029094.1 2 - 3 489 113
          NM_001163795.1 NP_001157267.1 2 - 3 501 113
          NM_001163796.1 NP_001157268.1 2 - 3 501 113
          NM_172623.2 NP_766211.2 2 - 3 501 113
          GenBank mRNA AK077228.1 - 2 - 3 501 113
          AK156085.1 - 2 - 3 504 113
          AY465530.1 - 2 - 3 459 113
          AY465531.1 - 2 - 3 459 113
          AY465532.1 - 2 - 3 459 113
          AY557629.1 - 2 - 3 437 113
          BC117091.1 - 2 - 3 476 40126410 113
          BC137666.1 - 2 - 3 476 8860685 113
          DQ087185.1 - 2 - 3 423 113
          DQ087186.1 - 2 - 3 423 113
          DQ186654.1 - 2 - 3 443 113
          DQ186655.1 - 2 - 3 455 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000934.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Treml4

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137616 AY557629.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137616 AK077228.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137616 AK156085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137616 NM_001163796.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137616 NM_172623.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204195 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 46677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98909 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180448 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169121 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149512 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137616 AY557629.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137616 AK077228.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137616 AK156085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137616 NM_001163796.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137616 NM_172623.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31216611 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149512 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180448 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03327817 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03327802 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169121 - -
          Whole Mouse Genome G4122A A_51_P468558 AK077228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468558 AK156085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468558 NM_001163796.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468558 NM_172623.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P468558 NM_001163795.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110036 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149512 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180448 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169121 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13338608 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13121366 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13121379 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13121367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13121361 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13121363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13121371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13236085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13121360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13533969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13121371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13356001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13355999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13338609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13533973 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445766 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445765 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445760 - -
          Affymetrix HT MG-430 PM Array Plate 1460014_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445758 - -
          Mouse Genome 430 2.0 Array 1460014_PM_at - -
          Mouse Genome 430 2.0 Array 1460014_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224840_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224840_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700008990.mm.1 NM_001033922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008990.mm.1 NM_001033922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008990.mm.1 NM_001163795.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008990.mm.1 NM_001163795.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008990.mm.1 NM_001163796.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008990.mm.1 NM_001163796.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008990.mm.1 NM_172623.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008990.mm.1 NM_172623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008990.mm.1 TC1700000934.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008993.mm.1 NM_001033922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008993.mm.1 NM_001033922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008993.mm.1 NM_001163795.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008993.mm.1 NM_001163795.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008993.mm.1 NM_001163796.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008993.mm.1 NM_001163796.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008993.mm.1 NM_172623.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008993.mm.1 NM_172623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008993.mm.1 TC1700000934.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008987.mm.1 NM_001033922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008987.mm.1 NM_001033922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008987.mm.1 NM_001163795.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008987.mm.1 NM_001163795.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008987.mm.1 NM_001163796.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008987.mm.1 NM_001163796.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008987.mm.1 NM_172623.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008987.mm.1 NM_172623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008987.mm.1 TC1700000934.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008991.mm.1 NM_001033922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008991.mm.1 NM_001033922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008991.mm.1 NM_001163795.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008991.mm.1 NM_001163795.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008991.mm.1 NM_001163796.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008991.mm.1 NM_001163796.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008991.mm.1 NM_172623.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008991.mm.1 NM_172623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008991.mm.1 TC1700000934.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008994.mm.1 NM_001033922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008994.mm.1 NM_001033922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008994.mm.1 NM_001163795.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008994.mm.1 NM_001163795.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008994.mm.1 NM_001163796.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008994.mm.1 NM_001163796.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008994.mm.1 NM_172623.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008994.mm.1 NM_172623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008994.mm.1 TC1700000934.mm.1 MTA TC
          Mouse Expression Set 430 1460014_PM_at - -
          Mouse Expression Set 430 1460014_at - -
          Affymetrix HT Mouse Genome 430B Array 1460014_at - -
          Affymetrix HT Mouse Genome 430B Array 1460014_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 506800 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5130735 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005130735 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2438678 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2438678 - -
          mouse-6 v1.1 (Illumina) scl50638.6.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 5130735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50638.6.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2438678 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3121765 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3121765 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2438678 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline