| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400011035.mm.1 |
NM_172701.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400011035.mm.1 |
NM_172701.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400011035.mm.1 |
TC0400001451.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400011038.mm.1 |
NM_172701.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400011038.mm.1 |
NM_172701.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400011038.mm.1 |
TC0400001451.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400011036.mm.1 |
NM_172701.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400011036.mm.1 |
NM_172701.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400011036.mm.1 |
TC0400001451.mm.1 |
MTA TC |