Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00555181_m1
          See other ANKLE1 GE Assays ›
          Gene Symbol:
          Ankle1
          Gene Name:
          ankyrin repeat and LEM domain containing 1
          Gene Aliases:
          8430438L13Rik, Ankrd41, C430003I23, Lem3
          Chromosome Location:
          Chr.8: 71406005 - 71410543 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00555181_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankle1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172756.2 NP_766344.2 6 - 7 1163 72
          GenBank mRNA AK049395.1 - 4 - 5 1470 72
          AK078830.1 - 5 - 6 1393 72
          AK080768.1 - 4 - 5 1476 72
          AK083552.1 - 4 - 5 1462 72
          AK087444.1 - 4 - 5 1481 72
          BC030436.1 - 6 - 7 1102 72
          BC094316.1 - 6 - 7 1120 72
          BC145659.1 - 2 - 3 386 72
          EU184014.1 - 6 - 7 1139 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000787.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ankle1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085412 BC094316.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085412 AK083552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085412 AK049395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085412 NM_172756.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030721 BC094316.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030721 AK083552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030721 AK087444.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030721 AK049395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030721 AK080768.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605532 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2635 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127416 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085412 BC094316.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085412 AK083552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085412 AK049395.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085412 NM_172756.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030721 BC094316.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030721 AK083552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030721 AK087444.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030721 AK049395.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030721 AK080768.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25956776 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127416 - -
          Whole Mouse Genome G4122A A_52_P220370 EU184014.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220370 BC030436.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220370 BC094316.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220370 BC145659.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P220370 AK083552.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127416 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10572608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572611 - -
          Affymetrix HT MG-430 PM Array Plate 1443978_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10572605 - -
          Mouse Genome 430 2.0 Array 1443978_at - -
          Mouse Genome 430 2.0 Array 1443978_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234396_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234396_at - -
          Affymetrix Mouse Clariom S Array PSR0800005662.mm.1 NM_172756.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005662.mm.1 TC0800000787.mm.1 MTA TC
          Mouse Expression Set 430 1443978_at - -
          Mouse Expression Set 430 1443978_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443978_at - -
          Affymetrix HT Mouse Genome 430B Array 1443978_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3840056 - -
          Illumina mouse-6 v1.0 expression beadchip 102650059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2723305 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2441304 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2723305 - -
          mouse-6 v1.1 (Illumina) scl33702.7_365-S - -
          mouse-6 v1.1 (Illumina) scl00234396.2_136-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840056 - -
          Illumina Sentrix mouse-6.v1 bead array 102650059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00234396.2_136-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840056 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102650059 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2441303 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2723305 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline