| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26350902 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26350901 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26350904 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26350903 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P68477 |
BC099418.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P68477 |
NM_172763.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P68477 |
BC099418.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P68477 |
NM_172763.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2047614 |
BC099418.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2047614 |
BY764604.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2047614 |
NR_028420.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2047614 |
AK044107.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2047614 |
AK049344.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
942768 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
304650 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
769590 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
53657 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
AK147278.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
AK147483.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
AK147597.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
AK158131.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
BC061219.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
BC079910.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
M84756.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
NM_001110783.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
NM_031158.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
U73972.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
X69063.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
X69064.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891016 |
XM_001476775.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354339 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
892728 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
846416 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
84222 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
56631 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114061 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P172050 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P68477 |
BC099418.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P68477 |
NM_172763.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P68477 |
BC099418.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P68477 |
NM_172763.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2047614 |
BC099418.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2047614 |
BY764604.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2047614 |
NR_028420.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2047614 |
AK044107.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2047614 |
AK049344.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350907 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350893 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350889 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350899 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350900 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350895 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350891 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350904 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350903 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350901 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350897 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26350905 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06720775 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114061 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06720757 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06720765 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P172050 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P68477 |
BC099418.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P68477 |
NM_172763.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P68477 |
BC099418.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P68477 |
NM_172763.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P466542 |
BC099418.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P466542 |
NR_028420.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P466542 |
AK044107.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P466542 |
AK049344.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P466542 |
AK089532.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P172050 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114061 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08119216 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07596556 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08119214 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26350904 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26350901 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26350902 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26350903 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350986 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26351001 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350880 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350854 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350948 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350856 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350922 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350952 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350940 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350949 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350872 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350972 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350954 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350848 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350851 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350857 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350958 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350946 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350863 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350912 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350930 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350907 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350918 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350870 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350968 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350850 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350894 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350891 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350911 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350943 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350883 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350860 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350956 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350992 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350862 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350900 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350846 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26351002 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350861 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350990 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350901 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350887 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350966 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350995 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350957 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350970 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350845 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350928 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350885 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350905 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350964 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350998 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350904 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350844 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350889 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26350898 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08119209 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350897 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08001272 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350850 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07954908 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350914 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350905 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08199651 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07596556 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07762618 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350857 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350891 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350875 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350889 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07489913 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350912 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08081149 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350846 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350852 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350870 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350915 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350848 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350887 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350885 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350854 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350883 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350856 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350862 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350894 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07971207 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350918 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350872 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07737036 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350900 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350901 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07909409 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350866 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350861 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07747471 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350893 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350860 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350922 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350904 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08129161 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26350844 |
- |
- |