Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00556259_m1
          See other RGP1 GE Assays ›
          Gene Symbol:
          Rgp1
          Gene Name:
          RGP1 retrograde golgi transport homolog (S. cerevisiae)
          Gene Aliases:
          1110029E03Rik, AI527196, F730001J03, mKIAA0258
          Chromosome Location:
          Chr.4: 43578735 - 43587487 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00556259_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172866.3 NP_766454.1 8 - 9 1236 80
          GenBank mRNA AK089288.1 - 8 - 9 1019 80
          AK129103.1 - 8 - 9 1022 80
          AK170117.1 - 8 - 9 1033 80
          BC086614.1 - 8 - 9 1042 30533256 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000442.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rgp1

          Gene Ontology Categories:

          Function(s) Process(es)

          retrograde transport, endosome to Golgi
          positive regulation of GTPase activity
          negative regulation of cellular protein catabolic process
          guanyl-nucleotide exchange factor activity
          Rab guanyl-nucleotide exchange factor activity
          Rab GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872582 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026458 AK089288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026458 AK129103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026458 NM_172866.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026458 AK170117.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026458 BC086614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063411 AK089288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063411 AK129103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063411 NM_172866.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063411 BC086614.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844390 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131957 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026458 AK089288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026458 AK129103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026458 NM_172866.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026458 AK170117.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026458 BC086614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063411 AK089288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063411 AK129103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063411 NM_172866.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063411 BC086614.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872581 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05217558 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131957 - -
          Whole Mouse Genome G4122A A_51_P385010 AK089288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385010 AK129103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385010 NM_172866.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385010 AK170117.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P385010 BC086614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343326 AK003952.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343326 AK129103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343326 NM_172866.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343326 BC086614.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102694 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104144 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131957 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03458646 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03458654 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03458664 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03458646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03286818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03448537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03458662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03576547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03286808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03366036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03782987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03250781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03866905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03458664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03334317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03497717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03286817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03286797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872665 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10504370 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504367 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504368 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504369 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504372 - -
          Affymetrix HT MG-430 PM Array Plate 1434338_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10504362 - -
          Affymetrix Mouse Gene 1.1 ST Array 10512594 - -
          Mouse Genome 430 2.0 Array 1434338_at - -
          Mouse Genome 430 2.0 Array 1434338_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242406_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242406_at - -
          Affymetrix Mouse Clariom S Array PSR0400003074.mm.1 NM_172866.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003074.mm.1 NM_172866.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003074.mm.1 TC0400000442.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400003068.mm.1 NM_172866.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003068.mm.1 NM_172866.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003068.mm.1 TC0400000442.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400003071.mm.1 NM_172866.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003071.mm.1 NM_172866.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003071.mm.1 TC0400000442.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400003072.mm.1 NM_172866.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003072.mm.1 NM_172866.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003072.mm.1 TC0400000442.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400003069.mm.1 NM_172866.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003069.mm.1 NM_172866.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003069.mm.1 TC0400000442.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400003070.mm.1 NM_172866.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003070.mm.1 NM_172866.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003070.mm.1 TC0400000442.mm.1 MTA TC
          Mouse Expression Set 430 1434338_at - -
          Mouse Expression Set 430 1434338_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434338_at - -
          Affymetrix HT Mouse Genome 430B Array 1434338_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 328439 - -
          ABI Mouse Genome Survey Microarray 584747 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1990086 - -
          Illumina mouse-6 v1.0 expression beadchip 101340161 - -
          Illumina mouse-6 v1.0 expression beadchip 1990086 - -
          Illumina mouse-6 v1.0 expression beadchip 1050139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001050139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710589 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2610190 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710589 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2610190 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2447658 - -
          mouse-6 v1.1 (Illumina) scl3210.1.1_13-S - -
          mouse-6 v1.1 (Illumina) scl0242406.9_36-S - -
          mouse-6 v1.1 (Illumina) scl00242406.1_148-S - -
          Illumina Sentrix mouse-6.v1 bead array 101340161 - -
          Illumina Sentrix mouse-6.v1 bead array 1990086 - -
          Illumina Sentrix mouse-6.v1 bead array 1050139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00242406.1_148-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0242406.9_36-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990086 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1050139 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101340161 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2610190 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710589 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710589 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2610190 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2447658 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline