Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00556326_m1
          See other CDCP2 GE Assays ›
          Gene Symbol:
          Cdcp2
          Gene Name:
          CUB domain containing protein 2
          Gene Aliases:
          D030010E02Rik
          Chromosome Location:
          Chr.4: 107096891 - 107114304 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00556326_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdcp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172873.3 NP_766461.1 2 - 3 1251 56
          GenBank mRNA AK050720.1 - 2 - 3 1252 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001121.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdcp2

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033116 AK050720.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033116 NM_172873.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385946 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603585 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13826 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104662 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134561 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033116 AK050720.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033116 NM_172873.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23188573 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104662 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134561 - -
          Whole Mouse Genome G4122A A_51_P421551 AK050720.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P421551 NM_172873.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119690 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114374 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104662 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04027077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04093640 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03397243 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04093639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23188592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04093639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04045580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03243671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03308137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03201870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03243659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03423718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04039067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03683517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03223438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03393644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03223432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03223419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03837849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03683515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03734818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03821373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03201874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03734809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03423717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03821374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03990902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04027078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03243649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03620973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03223428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03734811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03243658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03607391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03201877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04093640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03720015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03393645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04027077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23188527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03243655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03201875 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10506640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10506641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10506639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10506642 - -
          Affymetrix Mouse Gene 1.1 ST Array 10506638 - -
          Affymetrix Mouse Clariom S Array PSR0400007740.mm.1 NM_172873.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007740.mm.1 NM_172873.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400007740.mm.1 TC0400001121.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 360365 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2260438 - -
          Illumina mouse-6 v1.0 expression beadchip 2260438 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002260438 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251133 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251133 - -
          mouse-6 v1.1 (Illumina) scl25166.4.1_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 2260438 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25166.4.1_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260438 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251133 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251133 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline