| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600004437.mm.1 |
NM_172888.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004437.mm.1 |
NM_172888.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004437.mm.1 |
TC0600000576.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600004438.mm.1 |
NM_172888.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004438.mm.1 |
NM_172888.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004438.mm.1 |
TC0600000576.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600004439.mm.1 |
NM_172888.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004439.mm.1 |
NM_172888.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004439.mm.1 |
TC0600000576.mm.1 |
MTA TC |