Search
Search
| Gene Symbol: Fsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172904.2 | NP_766492.2 | 10 - 11 | 1769 | 64 | |
| GenBank mRNA | AK036674.1 | - | 1735 | 64 | ||
| AK052866.1 | - | 4 - 5 | 569 | 64 | ||
Set Membership: |
|