Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00557205_m1
          See other SERPINA3B GE Assays ›
          Gene Symbol:
          Serpina3b
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade A, member 3B
          Gene Aliases:
          6A1, A030003A19Rik
          Chromosome Location:
          Chr.12: 104127531 - 104139546 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00557205_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173024.3 NP_766612.1 4 - 5 1152 82
          GenBank mRNA AK037160.1 - 4 - 5 1154 82
          BC125418.1 - 4 - 5 1135 40129933 82
          BC145808.1 - 4 - 5 1135 40131111 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001039.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpina3b

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161450 AK037160.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161450 NM_173024.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735370 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36592 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151004 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159414 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161450 AK037160.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161450 NM_173024.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175442 AK037160.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175442 BC125418.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175442 BC145808.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175442 NM_173024.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28701611 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151004 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07733859 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159414 - -
          Whole Mouse Genome G4122A A_51_P175442 AK037160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175442 BC125418.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175442 BC145808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175442 NM_173024.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129144 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151004 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10030982 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10030984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28701694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09893893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10101887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10070318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09900123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09893900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09931639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09951552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10010753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10070314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10010757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10010754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09893903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10344727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10101888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10070317 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10398038 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398034 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398033 - -
          Affymetrix Mouse Gene 1.1 ST Array 10398032 - -
          Affymetrix Mouse Clariom S Array PSR1200007402.mm.1 NM_173024.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007402.mm.1 NM_173024.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007402.mm.1 TC1200001039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007400.mm.1 NM_173024.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007400.mm.1 NM_173024.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007400.mm.1 TC1200001039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007401.mm.1 NM_173024.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007401.mm.1 NM_173024.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007401.mm.1 TC1200001039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007403.mm.1 NM_173024.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007403.mm.1 NM_173024.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007403.mm.1 TC1200001039.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 498321 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5270010 - -
          Illumina mouse-6 v1.0 expression beadchip 5270010 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270010 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2611591 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2611591 - -
          mouse-6 v1.1 (Illumina) scl42836.5.1_252-S - -
          Illumina Sentrix mouse-6.v1 bead array 5270010 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42836.5.1_252-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270010 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2611591 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2611591 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline