Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00557206_m1
          See other ZBTB11 GE Assays ›
          Gene Symbol:
          Zbtb11
          Gene Name:
          zinc finger and BTB domain containing 11
          Gene Aliases:
          9230110G02Rik, ZNF-U69274
          Chromosome Location:
          Chr.16: 55972697 - 56008913 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00557206_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173026.2 NP_766614.2 4 - 5 1872 89
          GenBank mRNA AK033794.1 - 1 - 2 553 89
          AK048360.1 - 1 - 2 2153 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000738.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb11

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P256279 BC055403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P256279 NM_173026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P256279 BC055403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P256279 NM_173026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983828 BC055403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983828 AK033794.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983828 NM_173026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377299 AK048360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377299 AK033794.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377299 NM_173026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377299 AK048360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377299 AK033794.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377299 NM_173026.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125601 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115231 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120951 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983828 BC055403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983828 AK033794.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983828 NM_173026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P256279 BC055403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P256279 NM_173026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P256279 BC055403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P256279 NM_173026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377299 AK048360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377299 AK033794.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377299 NM_173026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377299 AK048360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377299 AK033794.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377299 NM_173026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P377299 AK048360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P377299 AK033794.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P377299 NM_173026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P377299 AK048360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P377299 AK033794.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P377299 NM_173026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P154005 BC055403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P154005 NM_173026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P256279 BC055403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P256279 NM_173026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P256279 BC055403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P256279 NM_173026.2 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12540428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12514369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12791654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12791650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12997589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13035601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12909800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12867320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12480387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30778770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10436250 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436249 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436248 - -
          Affymetrix HT MG-430 PM Array Plate 1454826_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10436239 - -
          Mouse Genome 430 2.0 Array 1454826_at - -
          Mouse Genome 430 2.0 Array 1454826_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 271377_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 271377_at - -
          Affymetrix Mouse Clariom S Array PSR1600006374.mm.1 NM_173026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006374.mm.1 NM_173026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006374.mm.1 TC1600000738.mm.1 MTA TC
          Mouse Expression Set 430 1454826_PM_at - -
          Mouse Expression Set 430 1454826_at - -
          Affymetrix HT Mouse Genome 430B Array 1454826_at - -
          Affymetrix HT Mouse Genome 430B Array 1454826_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 890999 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6520092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225321 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225321 - -
          mouse-6 v1.1 (Illumina) scl0271377.2_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0271377.2_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520092 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2504555 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline