Search
Search
| Gene Symbol: Cds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173370.3 | NP_775546.2 | 1 - 2 | 360 | 70 | |
| GenBank mRNA | AF533367.2 | - | 1 - 2 | 269 | 70 | |
| BC055292.1 | - | 1 - 2 | 360 | 6491596 | 70 | |
Set Membership: |
|