Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175087.4 | NP_780296.1 | 2 - 3 | 555 | 79 | |
GenBank mRNA | AK082699.1 | - | 2 - 3 | 556 | 79 | |
BC115586.1 | - | 2 - 3 | 556 | 40102369 | 79 | |
BC115587.1 | - | 2 - 3 | 555 | 79 | ||
DQ464060.1 | - | 2 - 3 | 556 | 79 | ||
DQ826418.1 | - | 2 - 3 | 547 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |