Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558325_m1
          See other TMEM127 GE Assays ›
          Gene Symbol:
          Tmem127
          Gene Name:
          transmembrane protein 127
          Gene Aliases:
          2310003P10Rik, AI314202, AI317350
          Chromosome Location:
          Chr.2: 127247975 - 127260764 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558325_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem127
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175145.3 NP_780354.1 3 - 4 587 65
          GenBank mRNA AK030324.1 - 3 - 4 608 65
          AK050216.1 - 3 - 4 557 65
          AK165880.1 - 3 - 4 654 65
          BC049832.1 - 3 - 4 587 6313802 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001910.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem127

          Gene Ontology Categories:

          Function(s) Process(es)

          endosome organization
          negative regulation of cell proliferation
          regulation of TOR signaling
          negative regulation of TOR signaling
          Rab GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612307 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612308 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612306 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612312 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612305 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612311 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 BC022794.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 BC049832.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 AK165880.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 AK033561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 NM_175145.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 BC022794.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 BC049832.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 AK165880.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 AK033561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P411019 NM_175145.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684574 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627411 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150777 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177150 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116276 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 BC022794.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 BC049832.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 AK165880.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 AK033561.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 NM_175145.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 BC022794.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 BC049832.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 AK165880.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 AK033561.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P411019 NM_175145.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21612334 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04684438 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177150 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116276 - -
          Whole Mouse Genome G4122A A_51_P411019 BC022794.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P411019 BC049832.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P411019 AK165880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P411019 AK033561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P411019 NM_175145.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P411019 BC022794.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P411019 BC049832.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P411019 AK165880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P411019 AK033561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P411019 NM_175145.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P282276 BC022794.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P282276 BC049832.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P282276 AK050216.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P282276 AK165880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P282276 AK030324.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126688 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116276 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02093272 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612307 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612305 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612308 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612312 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612311 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21612306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21612343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02093268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01882633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01533082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01882635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02093262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02093281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01882626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01882637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21612353 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10475769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475770 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10475771 - -
          Affymetrix HT MG-430 PM Array Plate 1433895_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433896_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10475767 - -
          Mouse Genome 430 2.0 Array 1433895_at - -
          Mouse Genome 430 2.0 Array 1433895_PM_at - -
          Mouse Genome 430 2.0 Array 1433896_at - -
          Mouse Genome 430 2.0 Array 1433896_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69470_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69470_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200014955.mm.1 NM_175145.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014955.mm.1 NM_175145.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014955.mm.1 TC0200001910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014956.mm.1 NM_175145.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014956.mm.1 NM_175145.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014956.mm.1 TC0200001910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014954.mm.1 NM_175145.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014954.mm.1 NM_175145.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014954.mm.1 TC0200001910.mm.1 MTA TC
          Mouse Expression Set 430 1433895_at - -
          Mouse Expression Set 430 1433896_at - -
          Mouse Expression Set 430 1433895_PM_at - -
          Mouse Expression Set 430 1433896_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433895_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433895_at - -
          Affymetrix HT Mouse Genome 430B Array 1433896_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433896_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 309120 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2940524 - -
          MouseWG-6 v2 Expression BeadChip 770341 - -
          Illumina mouse-6 v1.0 expression beadchip 2940524 - -
          Illumina mouse-6 v1.0 expression beadchip 770341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2746107 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222954 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222954 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2746107 - -
          mouse-6 v1.1 (Illumina) scl0003066.1_247-S - -
          mouse-6 v1.1 (Illumina) scl069470.6_81-S - -
          Illumina Sentrix mouse-6.v1 bead array 770341 - -
          Illumina Sentrix mouse-6.v1 bead array 2940524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003066.1_247-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl069470.6_81-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770341 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2594567 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2746107 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594567 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2746107 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl069470.6_81-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003066.1_247-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline