Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558327_s1
          See other ETD GE Assays ›
          Gene Symbol:
          Etd
          Gene Name:
          embryonic testis differentiation
          Gene Aliases:
          1700030K02Rik
          Chromosome Location:
          Chr.X: 53434918 - 53443576 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558327_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Etd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034074.1 2 311 95
          GenBank mRNA AK006555.1 - 2 272 95
          AY134667.1 - 2 311 95
          BC048661.1 - 2 6773570 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169327 BC048661.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169327 AY134667.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169327 NR_034074.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169327 AK006555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169327 BC048661.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169327 AY134667.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169327 NR_034074.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169327 AK006555.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536468 AJ414378.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536468 AK005241.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536468 AK010067.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536468 AK014118.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536468 AK043123.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536468 BC051534.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536468 BC060739.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536468 NM_025633.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33233 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107934 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169327 BC048661.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169327 AY134667.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169327 NR_034074.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169327 AK006555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169327 BC048661.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169327 AY134667.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169327 NR_034074.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169327 AK006555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169327 BC048661.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169327 AY134667.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169327 NR_034074.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169327 AK006555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169327 BC048661.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169327 AY134667.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169327 NR_034074.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169327 AK006555.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32387582 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03676260 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107934 - -
          Whole Mouse Genome G4122A A_51_P169327 BC048661.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169327 AY134667.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169327 NR_034074.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169327 AK006555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169327 BC048661.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169327 AY134667.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169327 NR_034074.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169327 AK006555.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115362 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132126 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14964172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14964185 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14964178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14964189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32387586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14641239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14947088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14964187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14824666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15020430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15020428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14964189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14641250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14866823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14737652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14799185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14927259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14992523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14964180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14964181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32387604 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10599651 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599653 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599652 - -
          Affymetrix Mouse Gene 1.1 ST Array 10599650 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 347451 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2570497 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570497 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731637 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731637 - -
          mouse-6 v1.1 (Illumina) scl54928.2.1_240-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570497 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54928.2.1_240-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570497 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2731637 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2731637 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0