Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558686_m1
          See other GPHB5 GE Assays ›
          Gene Symbol:
          Gphb5
          Gene Name:
          glycoprotein hormone beta 5
          Gene Aliases:
          OGH, Zlut1
          Chromosome Location:
          Chr.12: 75411720 - 75416781 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558686_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gphb5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175644.3 NP_783575.2 2 - 3 317 78
          GenBank mRNA AF467771.1 - 1 - 2 203 78
          AK078727.1 - 2 - 3 319 78
          BC111115.1 - 1 - 2 158 78
          BC139462.1 - 2 - 3 257 8862484 78
          BC139463.1 - 2 - 3 257 8862485 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001932.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gphb5

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of thyroid hormone mediated signaling pathway
          molecular_function
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058908 NM_175644.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058908 AK078727.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171102 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152860 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113038 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116214 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058908 NM_175644.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058908 AK078727.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P455528 NM_175644.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P455528 AK078727.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28548361 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113038 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116214 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152860 - -
          Whole Mouse Genome G4122A A_51_P455528 NM_175644.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P455528 AK078727.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126434 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116214 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152860 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113038 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116707 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116681 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116689 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116684 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116679 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10116704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28548363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10116677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10343014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10116685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10031218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09806771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09968806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10319994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10343002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10197028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10343007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10031221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10343022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09914573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10375110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10116678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28548382 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10401005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401006 - -
          Affymetrix Mouse Gene 1.1 ST Array 10401002 - -
          Affymetrix Mouse Clariom S Array PSR1200013270.mm.1 NM_175644.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013270.mm.1 NM_175644.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013270.mm.1 TC1200001932.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013271.mm.1 NM_175644.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013271.mm.1 NM_175644.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013271.mm.1 TC1200001932.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013272.mm.1 NM_175644.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013272.mm.1 NM_175644.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013272.mm.1 TC1200001932.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013269.mm.1 NM_175644.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013269.mm.1 NM_175644.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013269.mm.1 TC1200001932.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013273.mm.1 NM_175644.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013273.mm.1 NM_175644.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013273.mm.1 TC1200001932.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 610971 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2360446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245716 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245716 - -
          mouse-6 v1.1 (Illumina) scl42338.3.1_50-S - -
          Illumina Sentrix mouse-6.v1 bead array 2360446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42338.3.1_50-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245716 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245716 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline