Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558864_s1
          See other A530099J19RIK GE Assays ›
          Gene Symbol:
          A530099J19Rik
          Gene Name:
          RIKEN cDNA A530099J19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 19727417 - 19732955 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558864_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A530099J19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175688.4 NP_783619.2 2 759 108
          GenBank mRNA AK041317.1 - 2 760 108
          BC150908.1 - 2 638 9087819 108
          BC150909.1 - 2 638 9087820 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001525.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene A530099J19Rik

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974522 AK041317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974522 NM_175688.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360721 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156853 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974522 AK041317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974522 NM_175688.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P346015 AK041317.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P346015 NM_175688.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28863795 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156853 - -
          Whole Mouse Genome G4122A A_51_P346015 AK041317.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346015 NM_175688.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122849 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156853 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10712856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10712858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10712853 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10712825 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10712848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28863823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10712830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11054864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11054859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10712841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10712857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11054858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28863850 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10407984 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407983 - -
          Affymetrix Mouse Gene 1.1 ST Array 10407982 - -
          Affymetrix Mouse Clariom S Array PSR1300009685.mm.1 NM_175688.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009685.mm.1 NM_175688.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009685.mm.1 TC1300001525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009687.mm.1 NM_175688.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009687.mm.1 NM_175688.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009687.mm.1 TC1300001525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009688.mm.1 NM_175688.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009688.mm.1 NM_175688.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009688.mm.1 TC1300001525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009689.mm.1 NM_175688.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009689.mm.1 NM_175688.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009689.mm.1 TC1300001525.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 852852 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320347 - -
          Illumina mouse-6 v1.0 expression beadchip 2320347 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320347 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221040 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221040 - -
          mouse-6 v1.1 (Illumina) scl44247.2.1_245-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44247.2.1_245-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320347 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221040 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221040 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline