Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558877_m1
          See other PIANP GE Assays ›
          Gene Symbol:
          Pianp
          Gene Name:
          PILR alpha associated neural protein
          Gene Aliases:
          AI255183, AU067746, C530028O21Rik, Panp
          Chromosome Location:
          Chr.6: 124996720 - 125003096 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558877_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pianp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145926.1 NP_001139398.1 4 - 5 714 57
          NM_001145927.1 NP_001139399.1 4 - 5 698 57
          NM_175696.4 NP_783627.2 4 - 5 869 57
          GenBank mRNA AK082984.1 - 4 - 5 871 57
          AK148352.1 - 4 - 5 715 57
          BC065170.1 - 4 - 5 698 6413408 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001440.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pianp

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818073 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818074 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818072 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004532 BC065170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004532 NM_175696.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004532 AK082984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004532 AK188332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004532 NM_001145926.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759869 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118305 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004532 BC065170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004532 NM_175696.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004532 AK082984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004532 AK188332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004532 NM_001145926.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24818077 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118305 - -
          Whole Mouse Genome G4122A A_52_P563245 BC065170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563245 NM_175696.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563245 AK082984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563245 AK148352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563245 NM_001145926.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123769 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118305 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05130881 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05448155 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05448148 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05448141 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05130884 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05448170 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05448152 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05130885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05448147 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818073 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818072 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24818165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05853810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05448160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05025297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05130881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05448158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05410113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05448149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05025287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05448138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05130885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05731166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05731167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05448145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818087 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541757 - -
          Affymetrix HT MG-430 PM Array Plate 1455215_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1442066_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10541753 - -
          Mouse Genome 430 2.0 Array 1442066_at - -
          Mouse Genome 430 2.0 Array 1442066_PM_at - -
          Mouse Genome 430 2.0 Array 1455215_PM_at - -
          Mouse Genome 430 2.0 Array 1455215_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319352_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319352_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600011620.mm.1 TC0600001440.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011620.mm.1 NM_001145926.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011620.mm.1 NM_001145926.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011620.mm.1 NM_001145927.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011620.mm.1 NM_001145927.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011620.mm.1 NM_175696.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011620.mm.1 NM_175696.4 Refseq
          Mouse Expression Set 430 1442066_PM_at - -
          Mouse Expression Set 430 1442066_at - -
          Affymetrix HT Mouse Genome 430B Array 1442066_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442066_at - -
          Affymetrix HT Mouse Genome 430B Array 1455215_at - -
          Affymetrix HT Mouse Genome 430B Array 1455215_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 565093 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3710603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2642091 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2642091 - -
          mouse-6 v1.1 (Illumina) scl29513.8_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29513.8_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2642091 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2642091 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl29513.8_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline