Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00599710_m1
          See other NUDT1 GE Assays ›
          Gene Symbol:
          Nudt1
          Gene Name:
          nudix (nucleoside diphosphate linked moiety X)-type motif 1
          Gene Aliases:
          Mth1
          Chromosome Location:
          Chr.5: 140321627 - 140338137 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00599710_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nudt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008637.1 NP_032663.1 4 - 5 392 85
          GenBank mRNA AK011695.1 - 4 - 5 381 85
          AK088309.1 - 4 - 5 384 85
          AK168312.1 - 4 - 5 395 85
          BC098239.1 - 3 - 4 362 3469785 85
          D49956.1 - 4 - 5 392 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001689.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nudt1

          Panther Classification:

          Molecular Function -

          hydrolase phosphatase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleotide catabolic process
          dGTP catabolic process
          DNA repair
          DNA protection
          dATP catabolic process
          RNA binding
          8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
          hydrolase activity
          snoRNA binding
          8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity
          GTP diphosphatase activity
          metal ion binding
          ATP diphosphatase activity
          m7G(5')pppN diphosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24140928 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24140929 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085596 BC021940.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085596 BC098239.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085596 D49956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085596 NM_008637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085596 AK011695.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091681 AK088309.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091681 AK168312.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091681 BC098239.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091681 D49956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091681 NM_008637.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599863 AK039032.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599863 AK044233.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599863 XM_001478480.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599863 XM_484656.4 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649865 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103714 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091681 AK088309.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091681 AK168312.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091681 BC098239.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091681 D49956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091681 NM_008637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085596 BC021940.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085596 BC098239.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085596 D49956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085596 NM_008637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085596 AK011695.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435506 AK088309.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435506 AK168312.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435506 BC021940.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435506 BC098239.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435506 D49956.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140936 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103714 - -
          Whole Mouse Genome G4122A A_51_P435506 AK088309.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435506 AK168312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435506 BC021940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435506 BC098239.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435506 D49956.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103497 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103714 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04703691 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04701162 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04701165 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04701163 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24140928 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24140929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04438504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04661976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04438505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04193832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04122390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04122389 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526973 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526975 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526974 - -
          Affymetrix HT MG-430 PM Array Plate 1449349_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526972 - -
          Mouse Genome 430 2.0 Array 1449349_PM_at - -
          Mouse Genome 430 2.0 Array 1449349_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17766_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17766_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa17400_at - -
          Affymetrix Mouse Clariom S Array PSR0500015534.mm.1 TC0500001689.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015534.mm.1 NM_008637.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015534.mm.1 NM_008637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015533.mm.1 TC0500001689.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015533.mm.1 NM_008637.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015533.mm.1 NM_008637.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015535.mm.1 TC0500001689.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015535.mm.1 NM_008637.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015535.mm.1 NM_008637.1 Refseq
          Mouse Expression Set 430 1449349_PM_at - -
          Mouse Expression Set 430 1449349_at - -
          Murine Genome U74v2 Set 94372_at - -
          Murine Genome U74v2 Set 94372_PM_at - -
          Mouse Genome 430A 2.0 Array 1449349_PM_at - -
          Mouse Genome 430A 2.0 Array 1449349_at - -
          Affymetrix HT Mouse Genome 430A Array 1449349_at - -
          Affymetrix HT Mouse Genome 430A Array 1449349_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 906600 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2850600 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850600 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2688779 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2688779 - -
          mouse-6 v1.1 (Illumina) scl27052.5.1_64-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850600 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27052.5.1_64-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850600 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2880623 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2880623 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0