Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00599752_m1
          See other CST9 GE Assays ›
          Gene Symbol:
          Cst9
          Gene Name:
          cystatin 9
          Gene Aliases:
          M12, cresp, testatin
          Chromosome Location:
          Chr.2: 148835147 - 148838737 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00599752_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cst9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009979.1 NP_034109.1 1 - 2 279 107
          GenBank mRNA AB017157.1 - 1 - 2 254 107
          AK006429.1 - 1 - 2 271 107
          BC048486.1 - 1 - 2 268 6772736 107
          Y18243.1 - 1 - 2 279 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002203.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cst9

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          cysteine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 AB017157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 NM_009979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 AK006429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 BC048486.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 Y18243.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 AB017157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 NM_009979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 AK006429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 BC048486.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506297 Y18243.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969619 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150700 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 AB017157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 NM_009979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 AK006429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 BC048486.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 Y18243.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 AB017157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 NM_009979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 AK006429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 BC048486.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506297 Y18243.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 AB017157.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 NM_009979.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 AK006429.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 BC048486.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 Y18243.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 AB017157.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 NM_009979.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 AK006429.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 BC048486.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506297 Y18243.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21732448 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150700 - -
          Whole Mouse Genome G4122A A_51_P506297 AB017157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506297 NM_009979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506297 AK006429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506297 BC048486.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506297 Y18243.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506297 AB017157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506297 NM_009979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506297 AK006429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506297 BC048486.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506297 Y18243.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139071 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150700 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01575000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01965185 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01499130 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01965182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01965184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21732440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01450038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01577570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01368176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01602882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01602879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01602881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01965182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01575000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02179717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01965184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02179712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01577566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01499130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01577567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21732471 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10476926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476927 - -
          Affymetrix HT MG-430 PM Array Plate 1449285_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10476924 - -
          Mouse Genome 430 2.0 Array 1449285_PM_at - -
          Mouse Genome 430 2.0 Array 1449285_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13013_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13013_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200017431.mm.1 NM_009979.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017431.mm.1 NM_009979.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017431.mm.1 TC0200002203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017430.mm.1 NM_009979.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017430.mm.1 NM_009979.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017430.mm.1 TC0200002203.mm.1 MTA TC
          Mouse Expression Set 430 1449285_PM_at - -
          Mouse Expression Set 430 1449285_at - -
          Murine Genome U74v2 Set 103296_PM_at - -
          Murine Genome U74v2 Set 103296_at - -
          Mouse Genome 430A 2.0 Array 1449285_PM_at - -
          Mouse Genome 430A 2.0 Array 1449285_at - -
          Affymetrix HT Mouse Genome 430A Array 1449285_at - -
          Affymetrix HT Mouse Genome 430A Array 1449285_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2230706 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230706 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239816 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239816 - -
          mouse-6 v1.1 (Illumina) scl20152.3.1_18-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230706 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20152.3.1_18-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239816 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239816 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0