Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00600041_m1
          See other TRIP6 GE Assays ›
          Gene Symbol:
          Trip6
          Gene Name:
          thyroid hormone receptor interactor 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 137309899 - 137314487 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00600041_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trip6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011639.3 NP_035769.1 7 - 8 1362 65
          GenBank mRNA AF097511.1 - 7 - 8 1255 65
          AF116823.1 - 7 - 8 1357 65
          AK010435.1 - 7 - 8 1363 65
          AK146823.1 - 7 - 8 1312 65
          AK159387.1 - 7 - 8 1331 65
          AK166024.1 - 7 - 8 1526 65
          BC138685.1 - 7 - 8 1294 8861706 65
          BC138686.1 - 7 - 8 1294 8861707 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003441.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trip6

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cell adhesion
          release of cytoplasmic sequestered NF-kappaB
          positive regulation of cell migration
          interleukin-1 receptor binding
          protein binding
          zinc ion binding
          kinase binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AK159387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AK166024.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AF116823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AK010435.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AK146823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AK159387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AK166024.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AF116823.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AK010435.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P382914 AK146823.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764759 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166171 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AK159387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AK166024.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AF116823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AK010435.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AK146823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AK159387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AK166024.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AF116823.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AK010435.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P382914 AK146823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P121058 BC138686.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P121058 AK159387.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P121058 AK166024.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P121058 BC138685.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P121058 AF116823.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125139 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166171 - -
          Whole Mouse Genome G4122A A_52_P382914 AK159387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382914 AK166024.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382914 AF116823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382914 AK010435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382914 AK146823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382914 AK159387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382914 AK166024.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382914 AF116823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382914 AK010435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P382914 AK146823.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121058 BC138686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121058 AK159387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121058 AK166024.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121058 BC138685.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121058 AF116823.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123317 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135276 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166171 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04436145 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04436125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04877173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04225844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436149 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534724 - -
          Affymetrix HT MG-430 PM Array Plate 1449041_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534718 - -
          Mouse Genome 430 2.0 Array 1449041_PM_a_at - -
          Mouse Genome 430 2.0 Array 1449041_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22051_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22051_at - -
          Affymetrix Mouse Clariom S Array PSR0500031201.mm.1 TC0500003441.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031201.mm.1 NM_011639.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031201.mm.1 NM_011639.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031199.mm.1 TC0500003441.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031199.mm.1 NM_011639.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031199.mm.1 NM_011639.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031200.mm.1 TC0500003441.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031200.mm.1 NM_011639.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031200.mm.1 NM_011639.3 Refseq
          Mouse Expression Set 430 1449041_PM_a_at - -
          Mouse Expression Set 430 1449041_a_at - -
          Murine Genome U74v2 Set 94948_PM_at - -
          Murine Genome U74v2 Set 94948_at - -
          Murine Genome U74v2 Set 166413_PM_r_at - -
          Murine Genome U74v2 Set 166413_r_at - -
          Mouse Genome 430A 2.0 Array 1449041_a_at - -
          Mouse Genome 430A 2.0 Array 1449041_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449041_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449041_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6520609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520609 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1235489 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235489 - -
          mouse-6 v1.1 (Illumina) scl25882.4_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520609 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25882.4_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520609 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3009463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3009469 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3009463 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3009469 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline