Search
Search

Zfp385a| Gene Symbol: Zfp385a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013866.2 | NP_038894.2 | 4 - 5 | 449 | 79 | |
| GenBank mRNA | AF118566.1 | - | 4 - 5 | 447 | 79 | |
| AK137748.1 | - | 4 - 5 | 456 | 79 | ||
| BC017644.1 | - | 4 - 5 | 506 | 4504518 | 79 | |
Set Membership: |
|