Search
Search
| Gene Symbol: Ky | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024291.3 | NP_077253.3 | 1 - 2 | 144 | 56 | |
| GenBank mRNA | AJ293727.2 | - | 1 - 2 | 166 | 56 | |
| AK047076.1 | - | 1 - 2 | 535 | 56 | ||
Set Membership: |
|