Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00600419_m1
          See other LYAR GE Assays ›
          Gene Symbol:
          Lyar
          Gene Name:
          Ly1 antibody reactive clone
          Gene Aliases:
          MLZ-264
          Chromosome Location:
          Chr.5: 38220482 - 38234306 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00600419_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyar
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025281.3 NP_079557.2 5 - 6 630 67
          GenBank mRNA AK006373.1 - 5 - 6 564 67
          AK076727.1 - 5 - 6 635 67
          BC116923.1 - 4 - 5 486 40126242 67
          BC116927.1 - 4 - 5 486 40126246 67
          S60885.1 - 5 - 6 577 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000434.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lyar

          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23618271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23618272 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23618269 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074982 CD776101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074982 NM_025281.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074982 AK006373.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942826 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69634 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074982 CD776101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074982 NM_025281.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074982 AK006373.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23618285 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05539814 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01078440 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01078429 - -
          Whole Mouse Genome G4122A A_51_P452189 AK006373.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135319 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113187 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01078440 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04205435 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04142485 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04142491 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04205437 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23618272 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23618269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23618271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23618328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04692265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04743891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04233454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04233445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04743899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04743894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04233448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04702021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04138735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04246850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04138737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04233451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23618311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04702026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10521560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521564 - -
          Affymetrix HT MG-430 PM Array Plate 1417511_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10521555 - -
          Mouse Genome 430 2.0 Array 1417511_PM_at - -
          Mouse Genome 430 2.0 Array 1417511_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17089_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17089_at - -
          Affymetrix Mouse Clariom S Array PSR0500004317.mm.1 TC0500000434.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004317.mm.1 NM_025281.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004317.mm.1 NM_025281.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004313.mm.1 TC0500000434.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004313.mm.1 NM_025281.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004313.mm.1 NM_025281.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004314.mm.1 TC0500000434.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004314.mm.1 NM_025281.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004314.mm.1 NM_025281.3 Refseq
          Mouse Expression Set 430 1417511_at - -
          Mouse Expression Set 430 1417511_PM_at - -
          Mouse Genome 430A 2.0 Array 1417511_at - -
          Mouse Genome 430A 2.0 Array 1417511_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417511_at - -
          Affymetrix HT Mouse Genome 430A Array 1417511_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 869810 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103870091 - -
          Illumina mouse-6 v1.0 expression beadchip 103870091 - -
          Illumina mouse-6 v1.0 expression beadchip 7000402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000402 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2651126 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214143 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2651126 - -
          mouse-6 v1.1 (Illumina) scl27877.10.4_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 103870091 - -
          Illumina Sentrix mouse-6.v1 bead array 7000402 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27877.10.4_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103870091 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2651126 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214143 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2651126 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27877.10.4_15-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline