Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00600510_s1
          See other SPZ1 GE Assays ›
          Gene Symbol:
          Spz1
          Gene Name:
          spermatogenic leucine zipper 1
          Gene Aliases:
          1700027M20Rik, AI429130
          Chromosome Location:
          Chr.13: 92574631 - 92576232 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00600510_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spz1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030237.3 NP_084513.3 1 1027 90
          GenBank mRNA AF271697.1 - 1 761 90
          BC050748.1 - 1 967 6770895 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002425.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spz1

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256500 BC050748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256500 AK006437.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256500 NM_030237.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256500 AF271697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256500 BC050748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256500 AK006437.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256500 NM_030237.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256500 AF271697.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874327 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125045 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256500 BC050748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256500 AK006437.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256500 NM_030237.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256500 AF271697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256500 BC050748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256500 AK006437.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256500 NM_030237.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256500 AF271697.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29224690 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125045 - -
          Whole Mouse Genome G4122A A_51_P256500 BC050748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256500 AK006437.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256500 NM_030237.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256500 AF271697.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256500 BC050748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256500 AK006437.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256500 NM_030237.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256500 AF271697.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132023 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10790842 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10484400 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10484414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10878692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10484416 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10484420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29224731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10484412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11120085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10484409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10838855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32925451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10669387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10721390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32925450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10847807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10659520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10964009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11008721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10659518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11008722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29224722 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10411080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10411079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10411081 - -
          Affymetrix HT MG-430 PM Array Plate 1450653_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10411078 - -
          Mouse Genome 430 2.0 Array 1450653_PM_at - -
          Mouse Genome 430 2.0 Array 1450653_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79401_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79401_at - -
          Affymetrix Mouse Clariom S Array PSR1300016170.mm.1 NM_030237.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016170.mm.1 NM_030237.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016170.mm.1 TC1300002425.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300016169.mm.1 NM_030237.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016169.mm.1 NM_030237.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016169.mm.1 TC1300002425.mm.1 MTA TC
          Mouse Expression Set 430 1450653_at - -
          Mouse Expression Set 430 1450653_PM_at - -
          Murine Genome U74v2 Set 170168_at - -
          Murine Genome U74v2 Set 170168_PM_at - -
          Mouse Genome 430A 2.0 Array 1450653_PM_at - -
          Mouse Genome 430A 2.0 Array 1450653_at - -
          Affymetrix HT Mouse Genome 430A Array 1450653_at - -
          Affymetrix HT Mouse Genome 430A Array 1450653_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 660881 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5360068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2653834 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2653834 - -
          mouse-6 v1.1 (Illumina) scl43691.2.229_147-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43691.2.229_147-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2653834 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2653834 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline