Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00600660_s1
          See other OLFR1311 GE Assays ›
          Gene Symbol:
          Olfr1311
          Gene Name:
          olfactory receptor 1311
          Gene Aliases:
          MOR245-21
          Chromosome Location:
          Chr.2: 112020914 - 112021852 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00600660_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1311
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146274.1 NP_666386.1 1 564 103
          GenBank mRNA BC127967.1 - 1 568 40135022 103

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1311

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046524 NM_146274.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046524 BC127967.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140251 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046524 NM_146274.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046524 BC127967.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192824 NM_146274.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192824 BC127967.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140251 - -
          Whole Mouse Genome G4122A A_51_P192824 NM_146274.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192824 BC127967.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121367 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21527104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02210778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02210755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02210774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02210749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02210754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02210747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21527106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02210767 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485808 - -
          Affymetrix Mouse Gene 1.1 ST Array 10485807 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 814697 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4010128 - -
          Illumina mouse-6 v1.0 expression beadchip 4010128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2607096 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2607096 - -
          mouse-6 v1.1 (Illumina) scl18872.1.1_104-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18872.1.1_104-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2607095 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2607095 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0