Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00600677_s1
          See other OLFR1023 GE Assays ›
          Gene Symbol:
          Olfr1023
          Gene Name:
          olfactory receptor 1023
          Gene Aliases:
          MOR196-3
          Chromosome Location:
          Chr.2: 85886802 - 85887737 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00600677_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1023
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146587.2 NP_666798.2 1 640 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001254.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1023

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127387 NM_146587.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P351152 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120311 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P351152 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127387 NM_146587.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P351152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21399804 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120311 - -
          Whole Mouse Genome G4122A A_51_P351152 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113144 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120311 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01258629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01258623 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01258605 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01258603 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01258622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01964147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01258615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02163380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01157835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399789 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473476 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473475 - -
          Affymetrix Mouse Clariom S Array PSR0200009965.mm.1 NM_146587.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009965.mm.1 NM_146587.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009965.mm.1 TC0200001254.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 387155 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6770369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770369 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775910 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775910 - -
          mouse-6 v1.1 (Illumina) scl4936.1.1_100-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770369 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4936.1.1_100-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770369 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2775910 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2775910 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline