Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00612543_m1
          See other NDUFB9 GE Assays ›
          Gene Symbol:
          Ndufb9
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9
          Gene Aliases:
          1190008J14Rik
          Chromosome Location:
          Chr.15: 58933810 - 58939489 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00612543_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufb9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023172.3 NP_075661.1 1 - 2 132 64
          GenBank mRNA AK002254.1 - 1 - 2 133 64
          AK003540.1 - 1 - 2 133 64
          AK004522.1 - 1 - 2 133 64
          BC031539.1 - 1 - 2 114 3979845 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000423.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufb9

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial electron transport, NADH to ubiquinone
          transport
          oxidation-reduction process
          mitophagy in response to mitochondrial depolarization
          molecular_function
          NADH dehydrogenase (ubiquinone) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30251032 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30251034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30251033 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 BC031539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 AK004522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 NM_023172.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 AJ300739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 AK002254.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 BC031539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 AK004522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 NM_023172.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 AJ300739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177552 AK002254.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22048 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113568 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 BC031539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 AK004522.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 NM_023172.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 AJ300739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 AK002254.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 BC031539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 AK004522.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 NM_023172.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 AJ300739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177552 AK002254.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 BC031539.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 AK004522.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 NM_023172.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 AJ300739.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 AK002254.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 BC031539.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 AK004522.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 NM_023172.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 AJ300739.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177552 AK002254.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30251048 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113568 - -
          Whole Mouse Genome G4122A A_51_P177552 BC031539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177552 AK004522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177552 NM_023172.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177552 AJ300739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177552 AK002254.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177552 BC031539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177552 AK004522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177552 NM_023172.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177552 AJ300739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P177552 AK002254.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111397 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113568 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11981701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11981698 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11981697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11981691 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30251032 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30251034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30251033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30251086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12172782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12172795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12063689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11861400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12172777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12172769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30251077 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10424343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424341 - -
          Affymetrix HT MG-430 PM Array Plate 1455934_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436803_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1452184_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10424340 - -
          Mouse Genome 430 2.0 Array 1455934_at - -
          Mouse Genome 430 2.0 Array 1455934_PM_at - -
          Mouse Genome 430 2.0 Array 1452184_at - -
          Mouse Genome 430 2.0 Array 1436803_PM_a_at - -
          Mouse Genome 430 2.0 Array 1452184_PM_at - -
          Mouse Genome 430 2.0 Array 1436803_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66218_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66218_at - -
          Affymetrix Mouse Clariom S Array PSR1500003003.mm.1 NM_023172.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003003.mm.1 NM_023172.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003003.mm.1 TC1500000423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500003004.mm.1 NM_023172.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003004.mm.1 NM_023172.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003004.mm.1 TC1500000423.mm.1 MTA TC
          Mouse Expression Set 430 1455934_PM_at - -
          Mouse Expression Set 430 1436803_a_at - -
          Mouse Expression Set 430 1455934_at - -
          Mouse Expression Set 430 1436803_PM_a_at - -
          Mouse Expression Set 430 1452184_PM_at - -
          Mouse Expression Set 430 1452184_at - -
          Murine Genome U74v2 Set 100079_PM_at - -
          Murine Genome U74v2 Set 100079_at - -
          Mouse Genome 430A 2.0 Array 1452184_PM_at - -
          Mouse Genome 430A 2.0 Array 1436803_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1455934_PM_at - -
          Mouse Genome 430A 2.0 Array 1452184_at - -
          Mouse Genome 430A 2.0 Array 1455934_at - -
          Mouse Genome 430A 2.0 Array 1436803_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452184_at - -
          Affymetrix HT Mouse Genome 430A Array 1436803_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455934_at - -
          Affymetrix HT Mouse Genome 430A Array 1452184_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436803_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455934_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 909902 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3780348 - -
          Illumina mouse-6 v1.0 expression beadchip 3780348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605004 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605004 - -
          mouse-6 v1.1 (Illumina) scl066218.3_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 3780348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066218.3_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605004 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605004 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066218.3_27-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline